Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03062463_g1
          See other VOM2R70 GE Assays ›
          Gene Symbol:
          Vom2r70
          Gene Name:
          vomeronasal 2 receptor, 70
          Gene Aliases:
          RGD1562647
          Chromosome Location:
          Chr.14: 308569 - 338157 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03062463_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom2r70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099477.1 NP_001092947.1 2 348 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P329868 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P360972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157350 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P360972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P329868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P329868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P360972 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P360972 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P329868 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P360972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774989 - -
          Affymetrix Rat Gene 1.0 ST Array 10774978 - -
          Affymetrix Rat Gene 1.0 ST Array 10774955 - -
          Affymetrix Rat Gene 1.0 ST Array 10774974 - -
          Affymetrix Rat Gene 1.0 ST Array 10774960 - -
          Affymetrix Rat Gene 1.0 ST Array 10774952 - -
          Affymetrix Rat Gene 1.0 ST Array 10774987 - -
          Affymetrix Rat Gene 1.0 ST Array 10774947 - -
          Affymetrix Rat Gene 1.0 ST Array 10774950 - -
          Affymetrix Rat Gene 1.0 ST Array 10774980 - -
          Affymetrix Rat Gene 1.0 ST Array 10774953 - -
          Affymetrix Rat Gene 1.0 ST Array 10774972 - -
          Affymetrix Rat Gene 1.0 ST Array 10774959 - -
          Affymetrix Rat Gene 1.0 ST Array 10770902 - -
          Affymetrix Rat Gene 1.0 ST Array 10774988 - -
          Affymetrix Rat Gene 1.0 ST Array 10774961 - -
          Affymetrix Rat Gene 1.0 ST Array 10774990 - -
          Affymetrix Rat Gene 1.0 ST Array 10774991 - -
          Affymetrix Rat Gene 1.0 ST Array 10774984 - -
          Affymetrix Rat Gene 1.0 ST Array 10774977 - -
          Affymetrix Rat Gene 1.0 ST Array 10774982 - -
          Affymetrix Rat Gene 1.0 ST Array 10770901 - -
          Affymetrix Rat Gene 1.0 ST Array 10770898 - -
          Affymetrix Rat Gene 1.0 ST Array 10770899 - -
          Affymetrix Rat Gene 1.0 ST Array 10774979 - -
          Affymetrix Rat Gene 1.0 ST Array 10770900 - -
          Affymetrix Rat Gene 1.0 ST Array 10774976 - -
          Affymetrix Rat Gene 1.0 ST Array 10770897 - -
          Affymetrix Rat Gene 1.1 ST Array 10774948 - -
          Affymetrix Rat Gene 1.1 ST Array 10774983 - -
          Affymetrix Rat Gene 1.1 ST Array 10774951 - -
          Affymetrix Rat Gene 1.1 ST Array 10770896 - -
          Affymetrix Rat Gene 1.1 ST Array 10774973 - -
          Affymetrix Rat Gene 1.1 ST Array 10774975 - -
          Affymetrix Rat Gene 1.1 ST Array 10774985 - -
          Affymetrix Rat Gene 1.1 ST Array 10774946 - -
          Affymetrix Rat Gene 1.1 ST Array 10774971 - -
          Affymetrix Rat Clariom S Assay PSR1400000034.rn.1 TC1400000003.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000024.rn.1 TC1400000003.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000025.rn.1 TC1400000003.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000028.rn.1 TC1400000003.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000032.rn.1 TC1400000003.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000031.rn.1 TC1400000003.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10774971 - -
          Affymetrix Rat Gene 1.0 ST Array 10770896 - -
          Affymetrix Rat Gene 1.0 ST Array 10774951 - -
          Affymetrix Rat Gene 1.0 ST Array 10774946 - -
          Affymetrix Rat Gene 1.0 ST Array 10774975 - -
          Affymetrix Rat Gene 1.0 ST Array 10774985 - -
          Affymetrix Rat Gene 1.0 ST Array 10774948 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040384 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365121 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline