Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03417056_m1
          See other VOM2R63 GE Assays ›
          Gene Symbol:
          Vom2r63
          Gene Name:
          vomeronasal 2 receptor, 63
          Gene Aliases:
          RGD1564022
          Chromosome Location:
          Chr.12: 21008035 - 21077539 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03417056_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom2r63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099499.1 NP_001092969.1 1663 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001274.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Inventoried

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P134155 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007382 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007384 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007384 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134155 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007384 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134155 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007382 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447708 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101175 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760930 - -
          Affymetrix Rat Gene 1.0 ST Array 10757307 - -
          Affymetrix Rat Gene 1.0 ST Array 10760928 - -
          Affymetrix Rat Gene 1.0 ST Array 10760924 - -
          Affymetrix Rat Gene 1.0 ST Array 10760931 - -
          Affymetrix Rat Gene 1.0 ST Array 10760927 - -
          Affymetrix Rat Gene 1.0 ST Array 10757308 - -
          Affymetrix Rat Gene 1.0 ST Array 10760929 - -
          Affymetrix Rat Gene 1.0 ST Array 10757310 - -
          Affymetrix Rat Gene 1.0 ST Array 10757330 - -
          Affymetrix Rat Gene 1.0 ST Array 10760926 - -
          Affymetrix Rat Gene 1.0 ST Array 10760932 - -
          Affymetrix Rat Gene 1.0 ST Array 10757309 - -
          Affymetrix Rat Gene 1.1 ST Array 10760925 - -
          Affymetrix Rat Gene 1.1 ST Array 10757328 - -
          Affymetrix Rat Gene 1.1 ST Array 10757305 - -
          Affymetrix Rat Clariom S Assay PSR1200010789.rn.1 TC1200001274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010807.rn.1 TC1200001274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010781.rn.1 TC1200001274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010788.rn.1 TC1200001274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010806.rn.1 TC1200001274.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010798.rn.1 TC1200001274.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10757328 - -
          Affymetrix Rat Gene 1.0 ST Array 10760925 - -
          Affymetrix Rat Gene 1.0 ST Array 10757305 - -
          Affymetrix Rat Gene 1.0 ST Array 10760923 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355489 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline