Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03417059_m1
          See other VOM2R61 GE Assays ›
          Gene Symbol:
          Vom2r61
          Gene Name:
          vomeronasal 2 receptor, 61
          Gene Aliases:
          RGD1562930
          Chromosome Location:
          Chr.12: 19930584 - 19939565 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03417059_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vom2r61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099508.1 NP_001092978.1 1299 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001261.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134267 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42052 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052175 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052175 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052175 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760929 - -
          Affymetrix Rat Gene 1.0 ST Array 10760910 - -
          Affymetrix Rat Gene 1.0 ST Array 10760924 - -
          Affymetrix Rat Gene 1.0 ST Array 10760928 - -
          Affymetrix Rat Gene 1.0 ST Array 10760926 - -
          Affymetrix Rat Gene 1.0 ST Array 10757330 - -
          Affymetrix Rat Gene 1.0 ST Array 10760908 - -
          Affymetrix Rat Gene 1.0 ST Array 10760912 - -
          Affymetrix Rat Gene 1.0 ST Array 10760909 - -
          Affymetrix Rat Gene 1.0 ST Array 10760907 - -
          Affymetrix Rat Gene 1.0 ST Array 10760932 - -
          Affymetrix Rat Gene 1.0 ST Array 10760911 - -
          Affymetrix Rat Gene 1.0 ST Array 10760931 - -
          Affymetrix Rat Gene 1.1 ST Array 10760925 - -
          Affymetrix Rat Gene 1.1 ST Array 10760923 - -
          Affymetrix Rat Gene 1.1 ST Array 10760906 - -
          Affymetrix Rat Gene 1.1 ST Array 10757328 - -
          Rat Genome 230 2.0 Array 1385047_x_at - -
          Rat Genome 230 2.0 Array 1393688_PM_at - -
          Rat Genome 230 2.0 Array 1385047_PM_x_at - -
          Rat Genome 230 2.0 Array 1393688_at - -
          Affymetrix Rat Clariom S Assay PSR1200010651.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010662.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010654.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010668.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010650.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010653.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010667.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010663.rn.1 TC1200001261.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200010669.rn.1 TC1200001261.rn.1 RTA TC
          Rat Expression Set 230 1385047_x_at - -
          Rat Expression Set 230 1393688_PM_at - -
          Rat Expression Set 230 1385047_PM_x_at - -
          Rat Expression Set 230 1393688_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760923 - -
          Affymetrix Rat Gene 1.0 ST Array 10760925 - -
          Affymetrix Rat Gene 1.0 ST Array 10760906 - -
          Rat Genome U34 Set rc_AI012782_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline