Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03648288_m1
          See other RGD1565059 GE Assays ›
          Gene Symbol:
          RGD1565059
          Gene Name:
          similar to hypothetical protein E130311K13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 154485135 - 154508904 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03648288_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1565059
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127562.1 NP_001121034.1 4 - 5 573 118
          XM_006232447.2 XP_006232509.1 4 - 5 533 118
          XM_006232448.2 XP_006232510.1 3 - 4 473 118
          XM_008761057.1 XP_008759279.1 3 - 4 712 118
          GenBank mRNA BC167078.1 - 4 - 5 573 7133153 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003787.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of ERK1 and ERK2 cascade

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P667524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74690 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667524 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P667524 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P667524 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P667524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823458 - -
          Affymetrix Rat Gene 1.0 ST Array 10823457 - -
          Affymetrix Rat Gene 1.0 ST Array 10823460 - -
          Affymetrix Rat Gene 1.0 ST Array 10823459 - -
          Affymetrix Rat Gene 1.1 ST Array 10823456 - -
          Affymetrix Rat Clariom S Assay PSR0200025365.rn.1 TC0200003787.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10823456 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22229720 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362740 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0