Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03648297_g1
          See other CTS8 GE Assays ›
          Gene Symbol:
          Cts8
          Gene Name:
          cathepsin 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 3385851 - 3397621 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03648297_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cts8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128216.1 NP_001121688.1 2 - 3 251 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000042.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cysteine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23313 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017829 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017829 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017829 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10793634 - -
          Affymetrix Rat Gene 1.0 ST Array 10793637 - -
          Affymetrix Rat Gene 1.0 ST Array 10793636 - -
          Affymetrix Rat Gene 1.0 ST Array 10793635 - -
          Affymetrix Rat Gene 1.0 ST Array 10793640 - -
          Affymetrix Rat Gene 1.0 ST Array 10793639 - -
          Affymetrix Rat Gene 1.0 ST Array 10793638 - -
          Affymetrix Rat Gene 1.1 ST Array 10793633 - -
          Affymetrix Rat Clariom S Assay PSR1700000246.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000247.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000250.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000251.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000252.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000249.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700000248.rn.1 TC1700000042.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10793633 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline