Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn04338541_m1
See other CREM GE Assays ›
Gene Symbol:
Crem
Gene Name:
cAMP responsive element modulator
Gene Aliases:
Icer
Chromosome Location:
Chr.17: 57021704 - 57090888 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn04338541_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
2,694.00
Your Price
Online offer:
2694.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Crem
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001110860.2
NP_001104330.1
9 - 10
1286
88
NM_001271102.1
NP_001258031.1
2 - 3
240
88
NM_001271246.1
NP_001258175.1
3 - 4
384
88
NM_001271247.1
NP_001258176.1
3 - 4
430
88
XM_006254049.2
XP_006254111.1
8 - 9
1244
88
XM_006254051.2
XP_006254113.1
8 - 9
1070
88
XM_006254052.2
XP_006254114.1
7 - 8
1402
88
XM_006254053.2
XP_006254115.1
8 - 9
1024
88
XM_006254054.2
XP_006254116.1
7 - 8
1124
88
XM_006254055.2
XP_006254117.1
7 - 8
988
88
XM_006254057.2
XP_006254119.1
8 - 9
1091
88
XM_006254058.2
XP_006254120.1
7 - 8
1055
88
XM_006254060.2
XP_006254122.1
6 - 7
1255
88
XM_006254063.2
XP_006254125.1
6 - 7
974
88
XM_006254064.2
XP_006254126.1
7 - 8
835
88
XM_006254066.2
XP_006254128.1
6 - 7
799
88
XM_006254069.2
XP_006254131.1
3 - 4
663
88
XM_008771691.1
XP_008769913.1
3 - 4
709
88
XM_008771692.1
XP_008769914.1
3 - 4
305
88
XM_008771693.1
XP_008769915.1
2 - 3
947
88
XM_008771694.1
XP_008769916.1
2 - 3
284
88
XM_008771695.1
XP_008769917.1
2 - 3
263
88
GenBank mRNA
AB031423.1
-
3 - 4
305
88
FM033648.1
-
3 - 4
420
88
FM089481.1
-
3 - 4
385
88
FQ219343.1
-
2 - 3
237
88
FQ220921.1
-
2 - 3
239
88
U04835.1
-
3 - 4
314
88
Z15158.1
-
7 - 8
879
88
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1700000812.rn.1
Set Membership:
Amplicon length between 86 and 100
3' Most
Recommended Assay
Rattus norvegicus
Inventoried
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
spermatogenesis
cAMP-mediated signaling
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1700005468.rn.1
TC1700000812.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1700005469.rn.1
TC1700000812.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline