Search
Search

cdk1| Gene Symbol: cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203577.1 | NP_988908.1 | 6 - 7 | 737 | 99 | |
| XM_012966262.1 | XP_012821716.1 | 6 - 7 | 773 | 99 | ||
| XM_012966263.2 | XP_012821717.1 | 6 - 7 | 745 | 99 | ||
| GenBank mRNA | BC061617.1 | - | 6 - 7 | 737 | 99 | |
| BC077651.1 | - | 6 - 7 | 709 | 99 | ||
| CR762071.2 | - | 5 - 6 | 673 | 99 | ||
Set Membership: |
|