Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_164645848_10
          See other RAD51D GT Assays ›
          SNP ID:
          rs145309168
          Gene
          RAD51D RAD51L3-RFFL
          Gene Name
          RAD51 paralog D
          RAD51L3-RFFL readthrough
          Set Membership:
          -
          Chromosome Location:
          Chr.17: 35101008 - 35101008 on Build GRCh38
          Polymorphism:
          A/T, Transversion substitution
          Context Sequence [VIC/FAM]:

          CTGCTCTGAGGTCCCCCAGGTCCCA[A/T]TGTCTACCATCTCCTGGAAACCTGT

          Assay ID C_164645848_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 602954

          Literature Links:

          RAD51D PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          T (0.00)
          (1.00)
          Caucasian - Not Available CEPH (CEU) - Not Available
          EAS
          T (0.00)
          (1.00)
          African American - Not Available YRI (Yoruba) - Not Available
          SAS
          T (0.00)
          (1.00)
          Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR
          T (0.00)
          (1.00)
          Japanese - Not Available JPT (Japanese) - Not Available
          EUR
          T (0.00)
          (1.00)
          AMR
          T (0.00)
          (1.00)
          RAD51D - RAD51 paralog D
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001142571.1 1248 Missense Mutation AAT,ATT N,I 331 NP_001136043.1
          NM_002878.3 1248 Missense Mutation AAT,ATT N,I 311 NP_002869.3
          NM_133629.2 1248 Missense Mutation AAT,ATT N,I 199 NP_598332.1
          RAD51L3-RFFL - RAD51L3-RFFL readthrough
          There are no transcripts associated with this gene.

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic DNA recombinase assembly
          telomere maintenance
          double-strand break repair via homologous recombination
          DNA synthesis involved in DNA repair
          strand displacement
          DNA repair
          nucleotide-excision repair
          mitotic recombination
          reciprocal meiotic recombination
          response to ionizing radiation
          interstrand cross-link repair
          strand invasion
          regulation of cell cycle
          recombinase activity
          four-way junction DNA binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          endodeoxyribonuclease activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          gamma-tubulin binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline