Search
Search

HMGN4AGAAGGGAGAGAAGCTTCCCAAAGG[C/G]AGAAAGGGGAAAGCAGATGCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| HMGN4 - high mobility group nucleosomal binding domain 4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_006353.2 | 403 | Silent Mutation | GGC,GGG | G,G 55 | NP_006344.1 | |
| XM_017010171.1 | 403 | Silent Mutation | GGC,GGG | G,G 55 | XP_016865660.1 | |
| XM_017010172.1 | 403 | Silent Mutation | GGC,GGG | G,G 55 | XP_016865661.1 | |
| XM_017010173.1 | 403 | Silent Mutation | GGC,GGG | G,G 55 | XP_016865662.1 | |