Search
Search

BHMG1
DMPK
SIX5CTCCTGGGCGGCGCCAGACTGCGGT[A/G]AGTTGGCCGGCGTGGGCCACCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHMG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| BHMG1 - basic helix-loop-helix and HMG-box containing 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| DMPK - dystrophia myotonica protein kinase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001081560.2 | 2422 | Silent Mutation | CTC,CTT | L,L 611 | NP_001075029.1 | |
| NM_001081562.2 | 2422 | Missense Mutation | TCA,TTA | S,L 610 | NP_001075031.1 | |
| NM_001081563.2 | 2422 | Silent Mutation | CTC,CTT | L,L 626 | NP_001075032.1 | |
| NM_001288764.1 | 2422 | Silent Mutation | CTC,CTT | L,L 642 | NP_001275693.1 | |
| NM_001288765.1 | 2422 | Missense Mutation | TCA,TTA | S,L 526 | NP_001275694.1 | |
| NM_001288766.1 | 2422 | UTR 3 | NP_001275695.1 | |||
| NM_004409.4 | 2422 | Silent Mutation | CTC,CTT | L,L 616 | NP_004400.4 | |
| SIX5 - SIX homeobox 5 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||