Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__25752962_30
          See other PCDHGA1 GT Assays ›
          SNP ID:
          rs17097231
          Gene
          PCDHGA1 PCDHGA2 PCDHGA3 PCDHGA4 PCDHGA5 PCDHGB1 PCDHGB2
          Gene Name
          protocadherin gamma subfamily A, 1
          protocadherin gamma subfamily A, 2
          protocadherin gamma subfamily A, 3
          protocadherin gamma subfamily A, 4
          protocadherin gamma subfamily A, 5
          protocadherin gamma subfamily B, 1
          protocadherin gamma subfamily B, 2
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.5: 141360212 - 141360212 on Build GRCh38
          Polymorphism:
          C/G, Transversion substitution
          Context Sequence [VIC/FAM]:

          TTGCCCTTCCTGTTGTCTTTGTTCC[C/G]CGGGGCTCTCCCAGTCCAGATCCGC

          Assay ID C__25752962_30
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 606288 MIM: 606289 MIM: 606290 MIM: 606291 MIM: 606292 MIM: 606299 MIM: 606300

          Literature Links:

          PCDHGA1 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          G (0.13)
          (0.87)
          Caucasian - Not Available CEPH (CEU)
          G (0.12)
          (0.88)
          EAS
          G (0.12)
          (0.88)
          African American - Not Available YRI (Yoruba)
          G (0.19)
          (0.81)
          SAS
          G (0.14)
          (0.86)
          Chinese - Not Available JPT (Japanese)
          G (0.16)
          (0.84)
          AFR
          G (0.16)
          (0.84)
          Japanese - Not Available CHB (Han Chinese)
          G (0.15)
          (0.85)
          EUR
          G (0.07)
          (0.93)
          AMR
          G (0.17)
          (0.83)
          PCDHGA1 - protocadherin gamma subfamily A, 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_018912.2 77 Intron NP_061735.1
          NM_031993.1 77 Intron NP_114382.1
          PCDHGA2 - protocadherin gamma subfamily A, 2
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_018915.3 77 Intron NP_061738.1
          NM_032009.2 77 Intron NP_114398.1
          PCDHGA3 - protocadherin gamma subfamily A, 3
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_018916.3 77 Intron NP_061739.2
          NM_032011.1 77 Intron NP_114400.1
          PCDHGA4 - protocadherin gamma subfamily A, 4
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_018917.3 77 Intron NP_061740.2
          NM_032053.2 77 Intron NP_114442.2
          PCDHGA5 - protocadherin gamma subfamily A, 5
          There are no transcripts associated with this gene.
          PCDHGB1 - protocadherin gamma subfamily B, 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_018922.2 77 Intron NP_061745.1
          NM_032095.1 77 Intron NP_115266.1
          PCDHGB2 - protocadherin gamma subfamily B, 2
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_018923.2 77 Missense Mutation CCC,CGC P,R 26 NP_061746.1
          NM_032096.1 77 Missense Mutation CCC,CGC P,R 26 NP_115267.1

          Back To Top

          More Information


          Set Membership:

          HapMap

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          endosome to lysosome transport
          lipid tube assembly involved in organelle fusion
          calcium ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline