Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__26063260_10
          See other ENDOV GT Assays ›
          SNP ID:
          rs9913006
          Gene
          ENDOV LOC100294362 MIR4730
          Gene Name
          endonuclease V
          uncharacterized LOC100294362
          microRNA 4730
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.17: 80417131 - 80417131 on Build GRCh38
          Polymorphism:
          G/T, Transversion substitution
          Context Sequence [VIC/FAM]:

          TAAAGTTCACTCTGGTTGGCATGGC[G/T]TCAAGGCCCGTCATGACCTGTCCCC

          Assay ID C__26063260_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          Literature Links:

          ENDOV PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          G (0.42)
          (0.58)
          Caucasian - Not Available CEPH (CEU)
          T (0.39)
          (0.61)
          EAS
          G (0.40)
          (0.60)
          African American - Not Available YRI (Yoruba)
          G (0.22)
          (0.78)
          SAS
          G (0.48)
          (0.52)
          Chinese - Not Available JPT (Japanese)
          G (0.33)
          (0.67)
          AFR
          G (0.26)
          (0.74)
          Japanese - Not Available CHB (Han Chinese)
          G (0.40)
          (0.60)
          EUR
          T (0.41)
          (0.59)
          AMR
          G (0.45)
          (0.55)
          ENDOV - endonuclease V
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001164637.2 Intron NP_001158109.1
          NM_001164638.1 Intron NP_001158110.1
          NM_173627.4 Intron NP_775898.2
          XM_005257244.3 Intron XP_005257301.1
          XM_005257246.3 Intron XP_005257303.1
          XM_006721837.3 Intron XP_006721900.1
          XM_011524655.1 Intron XP_011522957.1
          XM_011524656.1 Intron XP_011522958.1
          XM_011524657.1 Intron XP_011522959.1
          XM_011524658.2 Intron XP_011522960.1
          XM_011524660.1 Intron XP_011522962.1
          XM_011524661.2 Intron XP_011522963.1
          XM_011524662.1 Intron XP_011522964.1
          XM_011524663.1 Intron XP_011522965.1
          XM_011524664.2 Intron XP_011522966.1
          XM_011524666.1 Intron XP_011522968.1
          XM_011524667.2 Intron XP_011522969.1
          XM_011524669.2 Intron XP_011522971.1
          XM_011524670.1 Intron XP_011522972.1
          XM_011524671.2 Intron XP_011522973.1
          XM_011524672.1 Intron XP_011522974.1
          XM_011524673.1 Intron XP_011522975.1
          XM_011524674.1 Intron XP_011522976.1
          XM_011524675.1 Intron XP_011522977.1
          XM_011524676.1 Intron XP_011522978.1
          XM_011524677.1 Intron XP_011522979.1
          XM_011524678.2 Intron XP_011522980.1
          XM_017024508.1 Intron XP_016879997.1
          XM_017024509.1 Intron XP_016879998.1
          XM_017024510.1 Intron XP_016879999.1
          XM_017024511.1 Intron XP_016880000.1
          XM_017024512.1 Intron XP_016880001.1
          XM_017024513.1 Intron XP_016880002.1
          XM_017024514.1 Intron XP_016880003.1
          XM_017024515.1 Intron XP_016880004.1
          XM_017024516.1 Intron XP_016880005.1
          XM_017024517.1 Intron XP_016880006.1
          XM_017024518.1 Intron XP_016880007.1
          XM_017024519.1 Intron XP_016880008.1
          XM_017024520.1 Intron XP_016880009.1
          LOC100294362 - uncharacterized LOC100294362
          There are no transcripts associated with this gene.
          MIR4730 - microRNA 4730
          There are no transcripts associated with this gene.

          Back To Top

          More Information


          Set Membership:

          HapMap

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          RNA phosphodiester bond hydrolysis, endonucleolytic
          magnesium ion binding
          single-stranded RNA binding
          endodeoxyribonuclease activity, producing 5'-phosphomonoesters
          endoribonuclease activity, producing 5'-phosphomonoesters
          structure-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline