Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__30633851_20
          See other DPYD GT Assays ›
          SNP ID:
          rs3918290
          Gene
          DPYD
          Gene Name
          dihydropyrimidine dehydrogenase
          Set Membership:
          > HapMap > DME > Validated > Inventoried
          Chromosome Location:
          Chr.1: 97450058 - 97450058 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          TGTTTTAGATGTTAAATCACACTTA[C/T]GTTGTCTGGAAAGTCAGCCTTTAGT

          Assay ID C__30633851_20
          Size VIC-FAM | 150 rxns
          Availability Inventoried
          Catalog # 4362691
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          43 submissions

          Phenotype:

          MIM: 612779

          Literature Links:

          DPYD PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          A (0.00)
          (1.00)
          Caucasian
          T (0.03)
          (0.97)
          CEPH (CEU)
          A (0.00)
          (1.00)
          EAS
          A (0.00)
          (1.00)
          African American
          T (0.00)
          (1.00)
          YRI (Yoruba) - Not Available
          SAS
          A (0.01)
          (0.99)
          Japanese
          T (0.00)
          (1.00)
          CHB (Han Chinese) - Not Available
          AFR
          A (0.00)
          (1.00)
          Chinese
          T (0.00)
          (1.00)
          JPT (Japanese) - Not Available
          EUR
          A (0.01)
          (0.99)
          AMR
          A (0.00)
          (1.00)
          DPYD - dihydropyrimidine dehydrogenase
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_000110.3 Intron NP_000101.2
          NM_001160301.1 Intron NP_001153773.1
          XM_005270562.3 Intron XP_005270619.2
          XM_006710397.3 Intron XP_006710460.1
          XM_017000507.1 Intron XP_016855996.1
          XM_017000508.1 Intron XP_016855997.1
          XM_017000509.1 Intron XP_016855998.1
          XM_017000510.1 Intron XP_016855999.1

          Back To Top

          More Information


          Set Membership:

          HapMap DME Validated Inventoried

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleobase catabolic process
          pyrimidine nucleobase catabolic process
          thymine catabolic process
          uracil catabolic process
          thymidine catabolic process
          beta-alanine biosynthetic process
          pyrimidine nucleoside catabolic process
          oxidation-reduction process
          protein binding
          dihydropyrimidine dehydrogenase (NADP+) activity
          protein homodimerization activity
          metal ion binding
          flavin adenine dinucleotide binding
          NADP binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-bxd7r:80/100.66.79.78:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline