Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__30634096_10
          See other SLC22A1 GT Assays ›
          SNP ID:
          rs12208357
          Gene
          SLC22A1
          Gene Name
          solute carrier family 22 member 1
          Set Membership:
          > DME > Validated > Inventoried
          Chromosome Location:
          Chr.6: 160122116 - 160122116 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          TCCTGGGGTGGCTGAGCTGAGCCAG[C/T]GCTGTGGCTGGAGCCCTGCGGAGGA

          Assay ID C__30634096_10
          Size VIC-FAM | 150 rxns
          Availability Inventoried
          Catalog # 4362691
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 602607

          Literature Links:

          SLC22A1 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          Applied Biosystems®
          Caucasian
          T (0.04)
          (0.96)
          African American
          T (0.00)
          (1.00)
          Japanese
          T (0.00)
          (1.00)
          Chinese
          T (0.00)
          (1.00)
          SLC22A1 - solute carrier family 22 member 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_003057.2 286 Missense Mutation CGC,TGC R,C 61 NP_003048.1
          NM_153187.1 286 Missense Mutation CGC,TGC R,C 61 NP_694857.1
          XM_005267102.4 328 Missense Mutation CGC,TGC R,C 61 XP_005267159.1
          XM_005267103.1 328 Missense Mutation CGC,TGC R,C 61 XP_005267160.1
          XM_005267104.4 Intron XP_005267161.1
          XM_005267105.4 Intron XP_005267162.1
          XM_006715552.1 328 Missense Mutation CGC,TGC R,C 61 XP_006715615.1
          XM_011536074.2 Intron XP_011534376.1

          Back To Top

          More Information


          Set Membership:

          DME Validated Inventoried

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          drug transmembrane transport
          establishment or maintenance of transmembrane electrochemical gradient
          organic cation transport
          quaternary ammonium group transport
          dopamine transport
          norepinephrine transport
          epinephrine transport
          protein homooligomerization
          ammonium transmembrane transport
          acetate ester transport
          acetylcholine transmembrane transporter activity
          dopamine transmembrane transporter activity
          norepinephrine transmembrane transporter activity
          protein binding
          secondary active organic cation transmembrane transporter activity
          organic cation transmembrane transporter activity
          quaternary ammonium group transmembrane transporter activity
          protein homodimerization activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch