Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__57466867_10
          See other KLHL17 GT Assays ›
          SNP ID:
          rs28561399
          Gene
          KLHL17 PERM1 PLEKHN1
          Gene Name
          kelch like family member 17
          PPARGC1 and ESRR induced regulator, muscle 1
          pleckstrin homology domain containing N1
          Set Membership:
          -
          Chromosome Location:
          Chr.1: 975093 - 975093 on Build GRCh38
          Polymorphism:
          A/G, Transition Substitution
          Context Sequence [VIC/FAM]:

          AGCACGGGAGACGTTGACACCACCG[A/G]ACGAGAAAGAAAAAAGAGAGAAGAG

          Assay ID C__57466867_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          32 submissions

          Phenotype:

          MIM: 615921

          Literature Links:

          KLHL17 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          A (0.17)
          (0.83)
          Caucasian - Not Available CEPH (CEU) - Not Available
          EAS
          A (0.08)
          (0.92)
          African American - Not Available YRI (Yoruba) - Not Available
          SAS
          A (0.25)
          (0.75)
          Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR
          A (0.25)
          (0.75)
          Japanese - Not Available JPT (Japanese) - Not Available
          EUR
          A (0.13)
          (0.87)
          AMR
          A (0.10)
          (0.90)
          KLHL17 - kelch like family member 17
          There are no transcripts associated with this gene.
          PERM1 - PPARGC1 and ESRR induced regulator, muscle 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001291366.1 2389 Intron NP_001278295.1
          NM_001291367.1 2389 Intron NP_001278296.1
          XM_017002583.1 2389 Intron XP_016858072.1
          XM_017002584.1 2389 Intron XP_016858073.1
          XM_017002585.1 2389 Intron XP_016858074.1
          PLEKHN1 - pleckstrin homology domain containing N1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001160184.1 2389 UTR 3 NP_001153656.1
          NM_032129.2 2389 UTR 3 NP_115505.2
          XM_006710944.3 2389 Intron XP_006711007.2
          XM_011542248.2 2389 Intron XP_011540550.2
          XM_017002474.1 2389 Intron XP_016857963.1
          XM_017002475.1 2389 Intron XP_016857964.1
          XM_017002476.1 2389 Intron XP_016857965.1
          XM_017002477.1 2389 Intron XP_016857966.1
          XM_017002478.1 2389 Intron XP_016857967.1
          XM_017002479.1 2389 Intron XP_016857968.1

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs74424855] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase transferase G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          response to muscle activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0