Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPR PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
| HNRNPR - heterogeneous nuclear ribonucleoprotein R | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001102397.2 | 2927 | Intron | NP_001095867.1 | |||
| NM_001102398.2 | 2927 | UTR 3 | NP_001095868.1 | |||
| NM_001102399.2 | 2927 | Intron | NP_001095869.1 | |||
| NM_001297620.1 | 2927 | UTR 3 | NP_001284549.1 | |||
| NM_001297621.1 | 2927 | Intron | NP_001284550.1 | |||
| NM_001297622.1 | 2927 | Intron | NP_001284551.1 | |||
| NM_005826.4 | 2927 | UTR 3 | NP_005817.1 | |||
| XM_005245711.4 | 2927 | Intron | XP_005245768.1 | |||
| XM_011540471.2 | 2927 | Intron | XP_011538773.1 | |||
| XM_011540472.2 | 2927 | Intron | XP_011538774.1 | |||
| XM_011540474.2 | 2927 | Intron | XP_011538776.1 | |||
| XM_011540475.2 | 2927 | Intron | XP_011538777.1 | |||
| XM_011540476.2 | 2927 | Intron | XP_011538778.1 | |||
| XM_011540477.2 | 2927 | Intron | XP_011538779.1 | |||
| XM_017000008.1 | 2927 | Intron | XP_016855497.1 | |||
| XM_017000009.1 | 2927 | Intron | XP_016855498.1 | |||
| XM_017000010.1 | 2927 | Intron | XP_016855499.1 | |||
Set Membership: |
|