Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___8240022_10
          See other MRAS GT Assays ›
          SNP ID:
          rs1564407
          Gene
          MRAS
          Gene Name
          muscle RAS oncogene homolog
          Set Membership:
          > HapMap > Validated
          Chromosome Location:
          Chr.3: 138352722 - 138352722 on Build GRCh38
          Polymorphism:
          G/A, Transition Substitution
          Context Sequence [VIC/FAM]:

          AAACAATATGATTTACATATGGGTT[G/A]TAGTGCTAGTAAGATTTTGGAGAGC

          Assay ID C___8240022_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 608435

          Literature Links:

          MRAS PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          C (0.38)
          (0.62)
          Caucasian
          G (0.31)
          (0.69)
          CEPH (CEU) - Not Available
          EAS
          C (0.27)
          (0.73)
          African American
          G (0.49)
          (0.51)
          YRI (Yoruba)
          T (0.34)
          (0.66)
          SAS
          C (0.29)
          (0.71)
          Chinese - Not Available JPT (Japanese)
          C (0.39)
          (0.61)
          AFR
          T (0.38)
          (0.62)
          Japanese - Not Available CHB (Han Chinese)
          C (0.16)
          (0.84)
          EUR
          C (0.33)
          (0.67)
          AMR
          C (0.25)
          (0.75)
          MRAS - muscle RAS oncogene homolog
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001085049.2 Intron NP_001078518.1
          NM_001252090.1 Intron NP_001239019.1
          NM_001252091.1 Intron NP_001239020.1
          NM_001252092.1 Intron NP_001239021.1
          NM_001252093.1 Intron NP_001239022.1
          NM_012219.4 Intron NP_036351.3
          XM_005247228.1 Intron XP_005247285.1
          XM_005247229.2 Intron XP_005247286.1
          XM_017005887.1 Intron XP_016861376.1

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs9289557] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          HapMap Validated

          Panther Classification:

          Molecular Function -

          G-protein enzyme modulator small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          Ras protein signal transduction
          multicellular organism development
          muscle organ development
          actin cytoskeleton organization
          GTPase activity
          protein binding
          GTP binding
          GTP-dependent protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-jmhj8:80/100.66.74.142:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline