Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___8870320_1_
          See other GRHL2 GT Assays ›
          SNP ID:
          rs1131862
          Gene
          GRHL2 NCALD
          Gene Name
          grainyhead like transcription factor 2
          neurocalcin delta
          Set Membership:
          > HapMap > Validated
          Chromosome Location:
          Chr.8: 101688100 - 101688100 on Build GRCh38
          Polymorphism:
          C/T, Transition Substitution
          Context Sequence [VIC/FAM]:

          ACCACTGGGAAGTTCCAAGAATGTA[C/T]GTGATGTTTTTTCGAGTCTGCAGAG

          Assay ID C___8870320_1_
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 608576 MIM: 606722

          Literature Links:

          GRHL2 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          G (0.24)
          (0.76)
          Caucasian
          C (0.20)
          (0.80)
          CEPH (CEU)
          G (0.27)
          (0.73)
          EAS
          G (0.25)
          (0.75)
          African American
          C (0.11)
          (0.89)
          YRI (Yoruba)
          G (0.14)
          (0.86)
          SAS
          G (0.35)
          (0.65)
          Chinese - Not Available JPT (Japanese)
          G (0.22)
          (0.78)
          AFR
          G (0.11)
          (0.89)
          Japanese - Not Available CHB (Han Chinese)
          G (0.23)
          (0.77)
          EUR
          G (0.23)
          (0.77)
          AMR
          G (0.35)
          (0.65)
          GRHL2 - grainyhead like transcription factor 2
          There are no transcripts associated with this gene.
          NCALD - neurocalcin delta
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001040624.1 2160 UTR 3 NP_001035714.1
          NM_001040625.1 2160 UTR 3 NP_001035715.1
          NM_001040626.1 2160 UTR 3 NP_001035716.1
          NM_001040627.1 2160 UTR 3 NP_001035717.1
          NM_001040628.1 2160 UTR 3 NP_001035718.1
          NM_001040629.1 2160 UTR 3 NP_001035719.1
          NM_001040630.1 2160 UTR 3 NP_001035720.1
          NM_032041.2 2160 UTR 3 NP_114430.2
          XM_011517332.2 2160 UTR 3 XP_011515634.1
          XM_011517333.2 2160 UTR 3 XP_011515635.1
          XM_011517334.2 2160 UTR 3 XP_011515636.1
          XM_011517335.2 2160 UTR 3 XP_011515637.1
          XM_017013900.1 2160 Intron XP_016869389.1
          XM_017013901.1 2160 UTR 3 XP_016869390.1
          XM_017013902.1 2160 UTR 3 XP_016869391.1

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs28362495] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          HapMap Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure
          vesicle-mediated transport
          calcium-mediated signaling
          ion transmembrane transport
          actin binding
          calcium ion binding
          protein binding
          ligand-gated ion channel activity
          tubulin binding
          clathrin binding
          alpha-tubulin binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0