Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001286
          Assay Name: hsa-miR-539
          miRBase Accession Number: MI0003514
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101047321 - 101047398 [+] on Build GRCh38
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Species: Human, Mouse, Rat, Bovine, Dog, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001286
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          GGAGAAAUUAUCCUUGGUGUGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-539-5p MIMAT0003163
          Mouse mmu-miR-539-5p MIMAT0003169
          Rat rno-miR-539-5p MIMAT0003176
          Bovine bta-miR-539 MIMAT0009342
          Dog cfa-miR-539 MIMAT0009909
          Pan troglodytes ptr-miR-539 MIMAT0008214
          View More Rows
          Pongo pygmaeus ppy-miR-539 MIMAT0015989
          Rhesus monkey mml-miR-539 MIMAT0006405

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-539
          Stem-loop Accession # MI0003514
          Stem-loop Sequence
          AUACUUGAGGAGAAAUUAUCCUUGGUGUGUUCGCUUUAUUUAUGAUGAAUCAUACAAGGACAAUUUCUUUUUGAGUAU
          Chromosome Location Chr. 14 - 101047321 - 101047398 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003163
          miRBase ID: hsa-miR-539-5p
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: Chr. 14 - 101047321 - 101047398 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-539
          Stem-loop Accession # MI0003520
          Stem-loop Sequence
          UACUUGAGGAGAAAUUAUCCUUGGUGUGUUGGCUCUUUUGGAUGAAUCAUACAAGGAUAAUUUCUUUUUGAGUA
          Chromosome Location Chr. 12 - 109728129 - 109728202 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003169
          miRBase ID: mmu-miR-539-5p
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: Chr. 12 - 109728129 - 109728202 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-539
          Stem-loop Accession # MI0003526
          Stem-loop Sequence
          AUACUUGAGGAGAAAUUAUCCUUGGUGUGUUUGCUCUUUUGGAUGAAUCAUACAAGGGUAAUUUCUUUUCGAGUAU
          Chromosome Location Chr. 6 - 133877832 - 133877907 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003176
          miRBase ID: rno-miR-539-5p
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: Chr. 6 - 133877832 - 133877907 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-539
          Stem-loop Accession # MI0009857
          Stem-loop Sequence
          AUACUUGAGGAGAAAUUAUCCUUGGUGUGUUCGCUUUAUUUGGGAUGAAUCAUACAAGGACAAUUUCUUUCUGAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009342
          miRBase ID: bta-miR-539
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-539
          Stem-loop Accession # MI0010408
          Stem-loop Sequence
          UACUUGAGGAGAAAUUAUCCUUGGUGUGUUCGCUGUGUUUGUGAGGAAUCAUACAAGGACAAUUUCUUUUUGAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009909
          miRBase ID: cfa-miR-539
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-539
          Stem-loop Accession # MI0008742
          Stem-loop Sequence
          UACUUGAGGAGAAAUUAUCCUUGGUGUGUUCGCUUUAUUUAUGAUGAAUCAUACAAGGACAAUUUCUUUUUGAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008214
          miRBase ID: ptr-miR-539
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-539
          Stem-loop Accession # MI0015038
          Stem-loop Sequence
          AUACUUGAGGAGAAAUUAUCCUUGGUGUGUUCGCUUUAUUUAUGAUGAAUCAUACAAGGACAAUUUCUUUUUGAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015989
          miRBase ID: ppy-miR-539
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-539
          Stem-loop Accession # MI0007815
          Stem-loop Sequence
          AUACUUGAGGAGAAAUUAUCCUUGGUGUGUUCGCUUUAUUUAUGAUGAAUCAUACAAGGACAAUUUCUUUUUGAGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006405
          miRBase ID: mml-miR-539
          Mature miRNA Sequence: GGAGAAAUUAUCCUUGGUGUGU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307662_pri
          TaqMan™ Pri-miRNA Assay : Rn03464852_pri
          TaqMan™ Pri-miRNA Assay : Hs03305121_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11336
          Ambion® Pre-miR™ miRNA Precursor : PM11336
          mirVana® miRNA inhibitor : MH11336
          mirVana® miRNA mimic : MC11336
          TaqMan™ Advanced miRNA Assay : rno481207_mir
          TaqMan™ Advanced miRNA Assay : mmu481207_mir
          TaqMan™ Advanced miRNA Assay : 478152_mir
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0


          miRBase Alias:

          Human: hsa-miR-539 (v17)
          Mouse: mmu-miR-539 (v15)
          Rat: rno-miR-539 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline