Search
Search

| Assay Name: | mmu-miR-384-3p |
|---|---|
| miRBase Accession Number: | MI0001146 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUUCCUAGAAAUUGUUCACAAU |
| Species: | Mouse, Rat, Cricetulus griseus, Dog, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000289, mir-384 |
Mature miRNA Sequence: | AUUCCUAGAAAUUGUUCACAAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-384-3p | MIMAT0001076 |
| Rat | rno-miR-384-3p | MIMAT0005310 |
| Cricetulus griseus | cgr-miR-384-3p | MIMAT0023949 |
| Dog | cfa-miR-384 | MIMAT0006752 |
| Rabbit | ocu-miR-384-3p | MIMAT0048432 |