Search
Search

| Assay Name: | mmu-miR-223-5p |
|---|---|
| miRBase Accession Number: | MI0000703 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGUGUAUUUGACAAGCUGAGUUG |
| Species: | Mouse, Rat, Dasypus novemcinctus, Guinea pig, Ornithorhynchus anatinus, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000067, mir-223 |
Mature miRNA Sequence: | CGUGUAUUUGACAAGCUGAGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-223-5p | MIMAT0017056 |
| Rat | rno-miR-223-5p | MIMAT0017165 |
| Dasypus novemcinctus | dno-miR-223-5p | MIMAT0047779 |
| Guinea pig | cpo-miR-223-5p | MIMAT0047134 |
| Ornithorhynchus anatinus | oan-miR-223-5p | MIMAT0007019 |
| Rabbit | ocu-miR-223-5p | MIMAT0048358 |