Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 479412_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-30c-1-3p
          Stem-loop Accession Number: MI0000736
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 40757284 - 40757372 [+] on Build GRCh38
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Species: Human, Mouse, Rat, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 479412_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          CUGGGAGAGGGUUGUUUACUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30c-1-3p MIMAT0004674
          Mouse mmu-miR-30c-1-3p MIMAT0004616
          Rat rno-miR-30c-1-3p MIMAT0004719
          Chrysemys picta cpi-miR-30c-3p MIMAT0037679
          Cricetulus griseus cgr-miR-30c MIMAT0041161
          Dasypus novemcinctus dno-miR-30c-3p MIMAT0047561
          View More Rows
          Rabbit ocu-miR-30c-3p MIMAT0048155

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30c-1
          Stem-loop Accession # MI0000736
          Stem-loop Sequence
          ACCAUGCUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA
          Chromosome Location Chr. 1 - 40757284 - 40757372 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004674
          miRBase ID: hsa-miR-30c-1-3p
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: Chr. 1 - 40757284 - 40757372 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-30c-1
          Stem-loop Accession # MI0000547
          Stem-loop Sequence
          ACCAUGUUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA
          Chromosome Location Chr. 4 - 120769534 - 120769622 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004616
          miRBase ID: mmu-miR-30c-1-3p
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: Chr. 4 - 120769534 - 120769622 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-30c-1
          Stem-loop Accession # MI0000866
          Stem-loop Sequence
          ACCAUGUUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA
          Chromosome Location Chr. 5 - 139699877 - 139699965 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004719
          miRBase ID: rno-miR-30c-1-3p
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: Chr. 5 - 139699877 - 139699965 [-] on Build Rnor_6.0

          Chrysemys picta

          Stem-loop ID cpi-mir-30c-1
          Stem-loop Accession # MI0029426
          Stem-loop Sequence
          GCCAUGCUGUAGCAUGUGUAAACAUCCUACACUCUCAGCUGUGGACUCAAGGUAGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGACUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037679
          miRBase ID: cpi-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-30c-1
          Stem-loop Accession # MI0033091
          Stem-loop Sequence
          ACCAUGUUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041161
          miRBase ID: cgr-miR-30c
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-30c-1
          Stem-loop Accession # MI0038936
          Stem-loop Sequence
          UGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047561
          miRBase ID: dno-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-30c-1
          Stem-loop Accession # MI0039249
          Stem-loop Sequence
          UGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048155
          miRBase ID: ocu-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAGGGUUGUUUACUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306692_pri
          TaqMan™ Pri-miRNA Assay : Rn03464776_pri
          TaqMan™ Pri-miRNA Assay : Hs03303371_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12472
          Ambion® Pre-miR™ miRNA Precursor : PM12472
          TaqMan™ MicroRNA Assay : 002108
          mirVana® miRNA inhibitor : MH12472
          mirVana® miRNA mimic : MC12472
          TaqMan™ Advanced miRNA Assay : mmu483230_mir
          TaqMan™ Advanced miRNA Assay : rno479412_mir
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-svw7g:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch