Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-638 |
---|---|
miRBase Accession Number: | MI0003653 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGGAUCGCGGGCGGGUGGCGGCCU |
Species: | Human |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000486, mir-638 |
Mature miRNA Sequence: | AGGGAUCGCGGGCGGGUGGCGGCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-638 | MIMAT0003308 |