Search
Search

| Assay Name: | hsa-miR-423-3p |
|---|---|
| miRBase Accession Number: | MI0001445 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCUCGGUCUGAGGCCCCUCAGU |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Goat, Guinea pig, Horse, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000329, mir-423 |
Mature miRNA Sequence: | AGCUCGGUCUGAGGCCCCUCAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-423-3p | MIMAT0001340 |
| Mouse | mmu-miR-423-3p | MIMAT0003454 |
| Rat | rno-miR-423-3p | MIMAT0005313 |
| Callithrix jacchus | cja-miR-423-3p | MIMAT0039484 |
| Cricetulus griseus | cgr-miR-423-3p | MIMAT0023957 |
| Dasypus novemcinctus | dno-miR-423-3p | MIMAT0047881 |
| View More Rows | ||
| Daubentonia madagascariensis | dma-miR-423 | MIMAT0049298 |
| Goat | chi-miR-423-3p | MIMAT0036221 |
| Guinea pig | cpo-miR-423-3p | MIMAT0047242 |
| Horse | eca-miR-423-3p | MIMAT0013040 |
| Nomascus leucogenys | nle-miR-423 | MIMAT0049358 |
| Otolemur garnettii | oga-miR-423 | MIMAT0049715 |
| Pan paniscus | ppa-miR-423 | MIMAT0049091 |
| Pan troglodytes | ptr-miR-423 | MIMAT0008142 |
| Pig | ssc-miR-423-3p | MIMAT0013881 |
| Pongo pygmaeus | ppy-miR-423-3p | MIMAT0015883 |
| Pteropus alecto | pal-miR-423-3p | MIMAT0039831 |
| Rabbit | ocu-miR-423-3p | MIMAT0048442 |
| Rhesus monkey | mml-miR-423-3p | MIMAT0006322 |
| Tupaia chinensis | tch-miR-423-3p | MIMAT0036525 |