Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno478327_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-423-3p
          Stem-loop Accession Number: MI0001445
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 30117079 - 30117172 [+] on Build GRCh38
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Species: Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Goat, Guinea pig, Horse, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno478327_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000329, mir-423

          Mature miRNA Sequence:

          AGCUCGGUCUGAGGCCCCUCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-423-3p MIMAT0001340
          Mouse mmu-miR-423-3p MIMAT0003454
          Rat rno-miR-423-3p MIMAT0005313
          Callithrix jacchus cja-miR-423-3p MIMAT0039484
          Cricetulus griseus cgr-miR-423-3p MIMAT0023957
          Dasypus novemcinctus dno-miR-423-3p MIMAT0047881
          View More Rows
          Daubentonia madagascariensis dma-miR-423 MIMAT0049298
          Goat chi-miR-423-3p MIMAT0036221
          Guinea pig cpo-miR-423-3p MIMAT0047242
          Horse eca-miR-423-3p MIMAT0013040
          Nomascus leucogenys nle-miR-423 MIMAT0049358
          Otolemur garnettii oga-miR-423 MIMAT0049715
          Pan paniscus ppa-miR-423 MIMAT0049091
          Pan troglodytes ptr-miR-423 MIMAT0008142
          Pig ssc-miR-423-3p MIMAT0013881
          Pongo pygmaeus ppy-miR-423-3p MIMAT0015883
          Pteropus alecto pal-miR-423-3p MIMAT0039831
          Rabbit ocu-miR-423-3p MIMAT0048442
          Rhesus monkey mml-miR-423-3p MIMAT0006322
          Tupaia chinensis tch-miR-423-3p MIMAT0036525

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-423
          Stem-loop Accession # MI0001445
          Stem-loop Sequence
          AUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUAACCCGCGC
          Chromosome Location Chr. 17 - 30117079 - 30117172 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001340
          miRBase ID: hsa-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: Chr. 17 - 30117079 - 30117172 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-423
          Stem-loop Accession # MI0004637
          Stem-loop Sequence
          ACUUGUGAGGAAAUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGCUUG
          Chromosome Location Chr. 11 - 77078064 - 77078172 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003454
          miRBase ID: mmu-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: Chr. 11 - 77078064 - 77078172 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-423
          Stem-loop Accession # MI0006145
          Stem-loop Sequence
          GAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCU
          Chromosome Location Chr. 10 - 63098366 - 63098444 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005313
          miRBase ID: rno-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: Chr. 10 - 63098366 - 63098444 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-423
          Stem-loop Accession # MI0032008
          Stem-loop Sequence
          CCGUACAUUUUCCAGGAUGGAAGCCCGAAGUUUGAGGGAGAAACUUGUGAGGAAAUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGCUUGAGUUUCUCCCCGCUUGGAUGCUCUCAGGGGCAGUGUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039484
          miRBase ID: cja-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-423
          Stem-loop Accession # MI0020523
          Stem-loop Sequence
          GAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023957
          miRBase ID: cgr-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-423
          Stem-loop Accession # MI0039105
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047881
          miRBase ID: dno-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-423
          Stem-loop Accession # MI0040033
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049298
          miRBase ID: dma-miR-423
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-423
          Stem-loop Accession # MI0030788
          Stem-loop Sequence
          ACUUGUGAGGAAAUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGCUUGAGUUUCUCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036221
          miRBase ID: chi-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-423
          Stem-loop Accession # MI0038769
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047242
          miRBase ID: cpo-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-423
          Stem-loop Accession # MI0012784
          Stem-loop Sequence
          AGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013040
          miRBase ID: eca-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-423
          Stem-loop Accession # MI0040095
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049358
          miRBase ID: nle-miR-423
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-423
          Stem-loop Accession # MI0040502
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049715
          miRBase ID: oga-miR-423
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-423
          Stem-loop Accession # MI0039800
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049091
          miRBase ID: ppa-miR-423
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-423
          Stem-loop Accession # MI0008663
          Stem-loop Sequence
          AUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008142
          miRBase ID: ptr-miR-423
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-423
          Stem-loop Accession # MI0013099
          Stem-loop Sequence
          AAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013881
          miRBase ID: ssc-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-423
          Stem-loop Accession # MI0014939
          Stem-loop Sequence
          AGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015883
          miRBase ID: ppy-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-423
          Stem-loop Accession # MI0032351
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039831
          miRBase ID: pal-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-423
          Stem-loop Accession # MI0039402
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048442
          miRBase ID: ocu-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-423
          Stem-loop Accession # MI0007739
          Stem-loop Sequence
          AUAAAGGAAGUUAGGCUGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGUCUUGCUUCCUACCCCGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006322
          miRBase ID: mml-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-423
          Stem-loop Accession # MI0031184
          Stem-loop Sequence
          UGAGGGGCAGAGAGCGAGACUUUUCUAUUUUCCAAAAGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036525
          miRBase ID: tch-miR-423-3p
          Mature miRNA Sequence: AGCUCGGUCUGAGGCCCCUCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307970_pri
          TaqMan™ Pri-miRNA Assay : Hs03303690_pri
          TaqMan™ Pri-miRNA Assay : Rn03465166_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13123
          Ambion® Pre-miR™ miRNA Precursor : PM13123
          TaqMan™ MicroRNA Assay : 002626
          mirVana® miRNA inhibitor : MH13123
          mirVana® miRNA mimic : MC13123
          TaqMan™ Advanced miRNA Assay : 478327_mir
          TaqMan™ Advanced miRNA Assay : mmu478327_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline