Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002248
          Assay Name: hsa-miR-142-5p
          miRBase Accession Number: MI0000458
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 58331232 - 58331318 [-] on Build GRCh38
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Species: Human, Mouse, Rat, Astatotilapia burtoni, Columba livia, Dasypus novemcinctus, Fugu rubripes, Gadus morhua, Guinea pig, Horse, Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pig, Pongo pygmaeus, Pundamilia nyererei, Rabbit, Rhesus monkey, Salmo salar, Tetraodon nigroviridis, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002248
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000084, mir-142

          Mature miRNA Sequence:

          CAUAAAGUAGAAAGCACUACU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-142-5p MIMAT0000433
          Mouse mmu-miR-142a-5p MIMAT0000154
          Rat rno-miR-142-5p MIMAT0000847
          Astatotilapia burtoni abu-miR-142-5p MIMAT0042103
          Columba livia cli-miR-142-5p MIMAT0038529
          Dasypus novemcinctus dno-miR-142-5p MIMAT0047657
          View More Rows
          Fugu rubripes fru-miR-142 MIMAT0002908
          Gadus morhua gmo-miR-142-5p MIMAT0044045
          Guinea pig cpo-miR-142-5p MIMAT0047008
          Horse eca-miR-142-5p MIMAT0013022
          Metriaclima zebra mze-miR-142 MIMAT0042338
          Neolamprologus brichardi nbr-miR-142 MIMAT0042531
          Oreochromis niloticus oni-miR-142 MIMAT0042701
          Ornithorhynchus anatinus oan-miR-142-5p MIMAT0006982
          Pig ssc-miR-142-5p MIMAT0013919
          Pongo pygmaeus ppy-miR-142-5p MIMAT0015765
          Pundamilia nyererei pny-miR-142 MIMAT0042917
          Rabbit ocu-miR-142-5p MIMAT0048245
          Rhesus monkey mml-miR-142-5p MIMAT0006199
          Salmo salar ssa-miR-142a-5p MIMAT0032361
          Tetraodon nigroviridis tni-miR-142a MIMAT0002909
          Zebrafish dre-miR-142a-5p MIMAT0001838

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-142
          Stem-loop Accession # MI0000458
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000433
          miRBase ID: hsa-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-142a
          Stem-loop Accession # MI0000167
          Stem-loop Sequence
          ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000154
          miRBase ID: mmu-miR-142a-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-142
          Stem-loop Accession # MI0000915
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000847
          miRBase ID: rno-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-142-1
          Stem-loop Accession # MI0033805
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042103
          miRBase ID: abu-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID abu-mir-142-2
          Stem-loop Accession # MI0033806
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042103
          miRBase ID: abu-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-142
          Stem-loop Accession # MI0029977
          Stem-loop Sequence
          GAGACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAAACAGCACUGCAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038529
          miRBase ID: cli-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-142
          Stem-loop Accession # MI0038991
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047657
          miRBase ID: dno-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-142
          Stem-loop Accession # MI0003223
          Stem-loop Sequence
          ACAGUGCUGUCACCCAUAAAGUAGAAAGCACUACUAAACUAUUCUACACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGCACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002908
          miRBase ID: fru-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-142-1
          Stem-loop Accession # MI0036183
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCCACAGUGUAGUGUUUCCUACUUUAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044045
          miRBase ID: gmo-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID gmo-mir-142-2
          Stem-loop Accession # MI0036000
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUUCAAUGCAAAGUGUAGUGUUUCCUACUUUAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044045
          miRBase ID: gmo-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-142
          Stem-loop Accession # MI0038649
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047008
          miRBase ID: cpo-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-142
          Stem-loop Accession # MI0012771
          Stem-loop Sequence
          ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013022
          miRBase ID: eca-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-142-1
          Stem-loop Accession # MI0034109
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042338
          miRBase ID: mze-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID mze-mir-142-2
          Stem-loop Accession # MI0034111
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042338
          miRBase ID: mze-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-142-1
          Stem-loop Accession # MI0034376
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042531
          miRBase ID: nbr-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID nbr-mir-142-2
          Stem-loop Accession # MI0034377
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042531
          miRBase ID: nbr-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-142-1
          Stem-loop Accession # MI0034610
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042701
          miRBase ID: oni-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID oni-mir-142-2
          Stem-loop Accession # MI0034611
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042701
          miRBase ID: oni-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-142
          Stem-loop Accession # MI0006772
          Stem-loop Sequence
          CGACAGACAGACAGACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAACAGCGCUGUAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUGGGCUUCGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006982
          miRBase ID: oan-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-142
          Stem-loop Accession # MI0013134
          Stem-loop Sequence
          GUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013919
          miRBase ID: ssc-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-142
          Stem-loop Accession # MI0014833
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015765
          miRBase ID: ppy-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-142-1
          Stem-loop Accession # MI0034893
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042917
          miRBase ID: pny-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID pny-mir-142-2
          Stem-loop Accession # MI0034895
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042917
          miRBase ID: pny-miR-142
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-142
          Stem-loop Accession # MI0039300
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048245
          miRBase ID: ocu-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-142
          Stem-loop Accession # MI0007631
          Stem-loop Sequence
          GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006199
          miRBase ID: mml-miR-142-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-142a-1
          Stem-loop Accession # MI0026500
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCACCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032361
          miRBase ID: ssa-miR-142a-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-142a-2
          Stem-loop Accession # MI0026501
          Stem-loop Sequence
          CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032361
          miRBase ID: ssa-miR-142a-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-142a
          Stem-loop Accession # MI0003224
          Stem-loop Sequence
          GUACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAAACUCUUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002909
          miRBase ID: tni-miR-142a
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-142a
          Stem-loop Accession # MI0002005
          Stem-loop Sequence
          CGUACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAAACCCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001838
          miRBase ID: dre-miR-142a-5p
          Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303162_pri
          TaqMan™ Pri-miRNA Assay : Rn03465064_pri
          TaqMan™ Pri-miRNA Assay : Mm03306331_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10979
          Ambion® Pre-miR™ miRNA Precursor : PM10979
          mirVana® miRNA inhibitor : MH10979
          mirVana® miRNA mimic : MC10979
          TaqMan™ Advanced miRNA Assay : 477911_mir
          TaqMan™ Advanced miRNA Assay : mmu481324_mir
          TaqMan™ Advanced miRNA Assay : rno481324_mir
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel


          miRBase Alias:

          Mouse: mmu-miR-142-5p (v20)
          Fugu rubripes: fru-miR-142a (v18)
          Ochrobactrum anthropi: oan-miR-142 (v18)
          Staphylococcus schleiferi: ssc-miR-142 (v16)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline