Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002467
          Assay Name: mmu-miR-193b
          miRBase Accession Number: MI0005484
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 13449523 - 13449601 [+] on Build GRCm38
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Species: Mouse, Alligator mississippiensis, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Goat, Guinea pig, Horse, Microcebus murinus, Monodelphis domestica, Ornithorhynchus anatinus, Otolemur garnettii, Pteropus alecto, Rabbit, Taeniopygia guttata, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002467
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000082, mir-193

          Mature miRNA Sequence:

          AACUGGCCCACAAAGUCCCGCU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-193b-3p MIMAT0004859
          Alligator mississippiensis ami-miR-193b-3p MIMAT0038211
          Chrysemys picta cpi-miR-193b-3p MIMAT0037811
          Columba livia cli-miR-193-3p MIMAT0038567
          Cricetulus griseus cgr-miR-193b-3p MIMAT0023826
          Dasypus novemcinctus dno-miR-193b-3p MIMAT0047724
          View More Rows
          Daubentonia madagascariensis dma-miR-193 MIMAT0049237
          Goat chi-miR-193b-3p MIMAT0036028
          Guinea pig cpo-miR-193b-3p MIMAT0047073
          Horse eca-miR-193b MIMAT0013056
          Microcebus murinus mmr-miR-193 MIMAT0049168
          Monodelphis domestica mdo-miR-193b-3p MIMAT0028653
          Ornithorhynchus anatinus oan-miR-193-3p MIMAT0006821
          Otolemur garnettii oga-miR-193b MIMAT0049662
          Pteropus alecto pal-miR-193b-3p MIMAT0039968
          Rabbit ocu-miR-193b-3p MIMAT0048306
          Taeniopygia guttata tgu-miR-193a-3p MIMAT0014585
          Tupaia chinensis tch-miR-193b-3p MIMAT0036532

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-193b
          Stem-loop Accession # MI0005484
          Stem-loop Sequence
          GGCCCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUGUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGGGUCA
          Chromosome Location Chr. 16 - 13449523 - 13449601 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004859
          miRBase ID: mmu-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: Chr. 16 - 13449523 - 13449601 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-193b
          Stem-loop Accession # MI0029776
          Stem-loop Sequence
          AAAGUUGUGGUUCCAGAGUCGGGGUUUUGGGGGCAAGAUGAGCUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGUGGUCACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038211
          miRBase ID: ami-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-193b
          Stem-loop Accession # MI0029515
          Stem-loop Sequence
          UUGUGGUUCCAGAGUCGGGGUUUUGGGGGCGAGAUGAGCUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGUGGUCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037811
          miRBase ID: cpi-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-193
          Stem-loop Accession # MI0029998
          Stem-loop Sequence
          GGUUCCAGAGUCGGGGUUUUGGGGGCGAGAUGAGCUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGUGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038567
          miRBase ID: cli-miR-193-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-193b
          Stem-loop Accession # MI0020439
          Stem-loop Sequence
          GCCCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUGUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGGGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023826
          miRBase ID: cgr-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-193b
          Stem-loop Accession # MI0039025
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047724
          miRBase ID: dno-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-193
          Stem-loop Accession # MI0039965
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049237
          miRBase ID: dma-miR-193
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-193b
          Stem-loop Accession # MI0030667
          Stem-loop Sequence
          GGAGGCUGUGGUCCCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGGGUCAUUCUAGACGGCGAGGGAUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036028
          miRBase ID: chi-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-193b
          Stem-loop Accession # MI0038682
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047073
          miRBase ID: cpo-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-193b
          Stem-loop Accession # MI0012800
          Stem-loop Sequence
          CAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013056
          miRBase ID: eca-miR-193b
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-193
          Stem-loop Accession # MI0039884
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049168
          miRBase ID: mmr-miR-193
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-193b
          Stem-loop Accession # MI0023850
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028653
          miRBase ID: mdo-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-193
          Stem-loop Accession # MI0006676
          Stem-loop Sequence
          GGUUGUGAUUCCAGAGUCGGGGUUUUGAGGGCGAGAUGAGUUUGUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGUGGUCACUUUGAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006821
          miRBase ID: oan-miR-193-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-193b
          Stem-loop Accession # MI0040445
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049662
          miRBase ID: oga-miR-193b
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-193b
          Stem-loop Accession # MI0032425
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039968
          miRBase ID: pal-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-193b
          Stem-loop Accession # MI0039331
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048306
          miRBase ID: ocu-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-193a
          Stem-loop Accession # MI0013813
          Stem-loop Sequence
          AGUCGGGGUUUUGGGGGCGAGAUGAGCUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGUGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014585
          miRBase ID: tgu-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-193b
          Stem-loop Accession # MI0031191
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036532
          miRBase ID: tch-miR-193b-3p
          Mature miRNA Sequence: AACUGGCCCACAAAGUCCCGCU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308440_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12596
          Ambion® Pre-miR™ miRNA Precursor : PM12596
          mirVana® miRNA inhibitor : MH12596
          mirVana® miRNA mimic : MC12596
          TaqMan™ Advanced miRNA Assay : mmu481599_mir
          FGMIR : TaqManTM Rodent MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0


          miRBase Alias:

          Mouse: mmu-miR-193b (v17)
          Ochrobactrum anthropi: oan-miR-193 (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-193a (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch