Search
Search

| Assay Name: | mmu-miR-381* |
|---|---|
| miRBase Accession Number: | MI0000798 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCGAGGUUGCCCUUUGUAUAUU |
| Species: | Mouse, Guinea pig, Pig, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGCGAGGUUGCCCUUUGUAUAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-381-5p | MIMAT0017081 |
| Guinea pig | cpo-miR-381-5p | MIMAT0047225 |
| Pig | ssc-miR-381-5p | MIMAT0017957 |
| Rhesus monkey | mml-miR-381-5p | MIMAT0026867 |