Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 483023_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-1246
          Stem-loop Accession Number: MI0006381
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 176600980 - 176601052 [-] on Build GRCh38
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Species: Human, Bovine, Pan troglodytes
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 483023_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000627, mir-1246

          Mature miRNA Sequence:

          AAUGGAUUUUUGGAGCAGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1246 MIMAT0005898
          Bovine bta-miR-1246 MIMAT0024567
          Pan troglodytes ptr-miR-1246 MIMAT0007972

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1246
          Stem-loop Accession # MI0006381
          Stem-loop Sequence
          UGUAUCCUUGAAUGGAUUUUUGGAGCAGGAGUGGACACCUGACCCAAAGGAAAUCAAUCCAUAGGCUAGCAAU
          Chromosome Location Chr. 2 - 176600980 - 176601052 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005898
          miRBase ID: hsa-miR-1246
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Chromosome Location: Chr. 2 - 176600980 - 176601052 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-1246
          Stem-loop Accession # MI0021112
          Stem-loop Sequence
          CAACAUAUUAAAUGGAUUUUUGGAGCAGGAAGUUGGAAUAGAGGCUUUCUCAGACAAAUACUUUACUGUGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024567
          miRBase ID: bta-miR-1246
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-1246
          Stem-loop Accession # MI0008452
          Stem-loop Sequence
          UGUAUCCUUGAAUGGAUUUUUGGAGCAGGAGUGGACACCUGACCCAAAGGAAAUCAAUCCAUAGGCUAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007972
          miRBase ID: ptr-miR-1246
          Mature miRNA Sequence: AAUGGAUUUUUGGAGCAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305724_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13182
          Ambion® Pre-miR™ miRNA Precursor : PM13182
          mirVana® miRNA inhibitor : MH13182
          mirVana® miRNA mimic : MC13182

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0