Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10149
          Assay Name: hsa-miR-363-3p
          miRBase Accession Number: MI0000764
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134169378 - 134169452 [-] on Build GRCh38
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Species: Human, Mouse, Callithrix jacchus, Cyprinus carpio, Dasypus novemcinctus, Guinea pig, Horse, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis, Xenopus laevis, Zebrafish
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10149
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000138, mir-363

          Mature miRNA Sequence:

          AAUUGCACGGUAUCCAUCUGUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-363-3p MIMAT0000707
          Mouse mmu-miR-363-3p MIMAT0000708
          Callithrix jacchus cja-miR-363 MIMAT0039359
          Cyprinus carpio ccr-miR-363 MIMAT0026293
          Dasypus novemcinctus dno-miR-363-3p MIMAT0047834
          Guinea pig cpo-miR-363-3p MIMAT0047193
          View More Rows
          Horse eca-miR-363 MIMAT0013212
          Pongo pygmaeus ppy-miR-363 MIMAT0015852
          Pteropus alecto pal-miR-363-3p MIMAT0039877
          Rabbit ocu-miR-363-3p MIMAT0048398
          Rhesus monkey mml-miR-363-3p MIMAT0006290
          Tupaia chinensis tch-miR-363-3p MIMAT0036586
          Xenopus laevis xla-miR-363-3p MIMAT0046589
          Zebrafish dre-miR-363-3p MIMAT0001874

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-363
          Stem-loop Accession # MI0000764
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUGAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location Chr. X - 134169378 - 134169452 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000707
          miRBase ID: hsa-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: Chr. X - 134169378 - 134169452 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-363
          Stem-loop Accession # MI0000765
          Stem-loop Sequence
          UGUUAUCAGGUGGAACACGAUGCAAUUUUGGUUGGUGUAAUAGGAGGAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location Chr. X - 52741693 - 52741767 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000708
          miRBase ID: mmu-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: Chr. X - 52741693 - 52741767 [-] on Build GRCm38

          Callithrix jacchus

          Stem-loop ID cja-mir-363
          Stem-loop Accession # MI0031873
          Stem-loop Sequence
          GUUCUGAUAUUUAGGCAUUGUAAAAUGAUCUGUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGACCUUUGUUGGCGACAUUCCUCAUCAGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039359
          miRBase ID: cja-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-363
          Stem-loop Accession # MI0023393
          Stem-loop Sequence
          UUUCGGGUGGAUGACCCCGCAAUUUUAAUAGUGGCAAAAAUCUUCCAACAAAAAUUGCACGGUAUCCAUCUGUAAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026293
          miRBase ID: ccr-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-363
          Stem-loop Accession # MI0039081
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047834
          miRBase ID: dno-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-363
          Stem-loop Accession # MI0038744
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGACUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047193
          miRBase ID: cpo-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-363
          Stem-loop Accession # MI0012958
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013212
          miRBase ID: eca-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-363
          Stem-loop Accession # MI0014910
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015852
          miRBase ID: ppy-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-363
          Stem-loop Accession # MI0032377
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039877
          miRBase ID: pal-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-363
          Stem-loop Accession # MI0039379
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUACAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048398
          miRBase ID: ocu-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-363
          Stem-loop Accession # MI0007709
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006290
          miRBase ID: mml-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-363
          Stem-loop Accession # MI0031252
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036586
          miRBase ID: tch-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-363-2
          Stem-loop Accession # MI0038327
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUAUUGAGCUUGGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046589
          miRBase ID: xla-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-363
          Stem-loop Accession # MI0002067
          Stem-loop Sequence
          UAAAUGCAAAUAUUUGUCUUGCUGUUUUCGGGUGGAUGACUCUGCAAUUUUAUUAGUGAUGGAAAAACUUCAAUAAAAAUUGCACGGUAUCCAUCUGUAAUCCGCUGGAUUCCAUUACCUGCGUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001874
          miRBase ID: dre-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295977_pri
          TaqMan™ Pri-miRNA Assay : Mm03307329_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10149
          TaqMan™ MicroRNA Assay : 001271
          mirVana® miRNA inhibitor : MH10149
          mirVana® miRNA mimic : MC10149
          TaqMan™ Advanced miRNA Assay : 478060_mir
          TaqMan™ Advanced miRNA Assay : mmu481384_mir


          miRBase Alias:

          Human: hsa-miR-363 (v17)
          Mouse: mmu-miR-363 (v15)
          Rat: rno-miR-363 (v15)
          Danio rerio: dre-miR-363 (v20)
          Macaca mulatta: mml-miR-363 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch