Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03307329_pri
          Assay Name:
          mmu-mir-363
          Stem-loop ID:
          mmu-mir-19b-2 mmu-mir-363 mmu-mir-92a-2
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.X: 52741983 - 52742066 [-] on Build GRCm38
          Stem-loop Location:
          Chr.X: 52741983 - 52742066 [-] on Build GRCm38 Chr.X: 52741693 - 52741767 [-] on Build GRCm38 Chr.X: 52741838 - 52741928 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-19b-3p mmu-miR-19b-2-5p mmu-miR-363-3p mmu-miR-363-5p mmu-miR-92a-3p mmu-miR-92a-2-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03307329_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          111

          Chromosome Location:

          Chr. X - 52741392 - 52741392 [+] on 52741392 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-19b-3p MIMAT0000513
          Mouse mmu-miR-19b-2-5p MIMAT0017010
          Mouse mmu-miR-363-3p MIMAT0000708
          Mouse mmu-miR-363-5p MIMAT0017076
          Mouse mmu-miR-92a-3p MIMAT0000539
          Mouse mmu-miR-92a-2-5p MIMAT0004635

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-19b-2
          Stem-loop Accession # MI0000546
          Stem-loop Sequence
          ACUUACGAUUAGUUUUGCAGAUUUGCAGUUCAGCGUAUAUGUGAAUAUAUGGCUGUGCAAAUCCAUGCAAAACUGAUUGUGGGA
          Chromosome Location Chr. X - 52741983 - 52742066 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000513
          miRBase ID: mmu-miR-19b-3p
          Mature miRNA Sequence: UGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location: Chr. X - 52741983 - 52742066 [-] on Build GRCm38

          miRBase Accession #: MIMAT0017010
          miRBase ID: mmu-miR-19b-2-5p
          Mature miRNA Sequence: AGUUUUGCAGAUUUGCAGUUCAGC
          Chromosome Location: Chr. X - 52741983 - 52742066 [-] on Build GRCm38

          Gene Family ID MIPF0000011, mir-19
          Distance to Amplicon 527 bases
          Stem-loop ID mmu-mir-363
          Stem-loop Accession # MI0000765
          Stem-loop Sequence
          UGUUAUCAGGUGGAACACGAUGCAAUUUUGGUUGGUGUAAUAGGAGGAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location Chr. X - 52741693 - 52741767 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000708
          miRBase ID: mmu-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: Chr. X - 52741693 - 52741767 [-] on Build GRCm38

          miRBase Accession #: MIMAT0017076
          miRBase ID: mmu-miR-363-5p
          Mature miRNA Sequence: CAGGUGGAACACGAUGCAAUUU
          Chromosome Location: Chr. X - 52741693 - 52741767 [-] on Build GRCm38

          Gene Family ID MIPF0000138, mir-363
          Distance to Amplicon 237 bases
          Stem-loop ID mmu-mir-92a-2
          Stem-loop Accession # MI0000580
          Stem-loop Sequence
          UGCCCAUUCAUCCACAGGUGGGGAUUGGUGGCAUUACUUGUGUUAGAUAUAAAGUAUUGCACUUGUCCCGGCCUGAGGAAGAAAGAGGGUU
          Chromosome Location Chr. X - 52741838 - 52741928 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000539
          miRBase ID: mmu-miR-92a-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: Chr. X - 52741838 - 52741928 [-] on Build GRCm38

          miRBase Accession #: MIMAT0004635
          miRBase ID: mmu-miR-92a-2-5p
          Mature miRNA Sequence: AGGUGGGGAUUGGUGGCAUUAC
          Chromosome Location: Chr. X - 52741838 - 52741928 [-] on Build GRCm38

          Gene Family ID MIPF0000013, mir-25
          Distance to Amplicon 382 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10149
          Ambion® Pre-miR™ miRNA Precursor : PM10149
          TaqMan™ MicroRNA Assay : 001271
          mirVana® miRNA inhibitor : MH10149
          mirVana® miRNA mimic : MC10149
          TaqMan™ Advanced miRNA Assay : 478060_mir
          TaqMan™ Advanced miRNA Assay : mmu481384_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19384
          Ambion® Pre-miR™ miRNA Precursor : PM19384
          TaqMan™ MicroRNA Assay : 463354_mat
          mirVana® miRNA inhibitor : MH19384
          mirVana® miRNA mimic : MC19384
          TaqMan™ Advanced miRNA Assay : mmu481820_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10312
          Ambion® Pre-miR™ miRNA Precursor : PM10312
          TaqMan™ MicroRNA Assay : 000430
          mirVana® miRNA inhibitor : MH10312
          mirVana® miRNA mimic : MC10312
          Ambion® Anti-miR™ miRNA Inhibitor : AM13021
          Ambion® Pre-miR™ miRNA Precursor : PM13021
          TaqMan™ MicroRNA Assay : 002496
          mirVana® miRNA inhibitor : MH13021
          mirVana® miRNA mimic : MC13021
          TaqMan™ Advanced miRNA Assay : mmu482577_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10629
          Ambion® Pre-miR™ miRNA Precursor : PM10629
          TaqMan™ MicroRNA Assay : 000396
          mirVana® miRNA inhibitor : MH10629
          mirVana® miRNA mimic : MC10629
          TaqMan™ Advanced miRNA Assay : 478264_mir
          TaqMan™ Advanced miRNA Assay : mmu478264_mir
          TaqMan™ Advanced miRNA Assay : rno478264_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20610
          Ambion® Pre-miR™ miRNA Precursor : PM20610
          TaqMan™ MicroRNA Assay : 464742_mat
          mirVana® miRNA inhibitor : MH20610
          mirVana® miRNA mimic : MC20610
          TaqMan™ Advanced miRNA Assay : mmu481635_mir


          miRBase Alias:

          Mouse: mmu-mir-92a-2 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch