Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10312
          Assay Name: mmu-miR-92a-3p
          miRBase Accession Number: MI0000719
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 115044427 - 115044506 [+] on Build GRCm38
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Species: Mouse, Rat, Chicken, Oryzias latipes, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10312

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000013, mir-25

          Mature miRNA Sequence:

          UAUUGCACUUGUCCCGGCCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-92a-3p MIMAT0000539
          Rat rno-miR-92a-3p MIMAT0000816
          Chicken gga-miR-92-3p MIMAT0001109
          Oryzias latipes ola-miR-92a MIMAT0022534
          Xenopus tropicalis xtr-miR-92a MIMAT0003580

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-92a-1
          Stem-loop Accession # MI0000719
          Stem-loop Sequence
          CUUUCUACACAGGUUGGGAUUUGUCGCAAUGCUGUGUUUCUCUGUAUGGUAUUGCACUUGUCCCGGCCUGUUGAGUUUGG
          Chromosome Location Chr. 14 - 115044427 - 115044506 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000539
          miRBase ID: mmu-miR-92a-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: Chr. X - 52741838 - 52741928 [-] on Build GRCm38

          Stem-loop ID mmu-mir-92a-2
          Stem-loop Accession # MI0000580
          Stem-loop Sequence
          UGCCCAUUCAUCCACAGGUGGGGAUUGGUGGCAUUACUUGUGUUAGAUAUAAAGUAUUGCACUUGUCCCGGCCUGAGGAAGAAAGAGGGUU
          Chromosome Location Chr. X - 52741838 - 52741928 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000539
          miRBase ID: mmu-miR-92a-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: Chr. X - 52741838 - 52741928 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-92a-1
          Stem-loop Accession # MI0000878
          Stem-loop Sequence
          CUUUCUACACAGGUUGGGAUUUGUCGCAAUGCUGUGUUUCUGUAUAGUAUUGCACUUGUCCCGGCCUGUUGAGUUUGG
          Chromosome Location Chr. 15 - 100180586 - 100180663 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000816
          miRBase ID: rno-miR-92a-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: Chr. 15 - 100180586 - 100180663 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-92a-2
          Stem-loop Accession # MI0000879
          Stem-loop Sequence
          UGCCCAUUCAUCCACAGGUGGGGAUUAGUGCCAUUACUUGUGUUAGAUAAAAAGUAUUGCACUUGUCCCGGCCUGAGGAAGAAAAGAGGGUU
          Chromosome Location Chr. X - 140117249 - 140117340 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000816
          miRBase ID: rno-miR-92a-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: Chr. 15 - 100180586 - 100180663 [+] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-mir-92-1
          Stem-loop Accession # MI0001179
          Stem-loop Sequence
          CUUUCUACACAGGUUGGGAUCAGUUGCAAUGCUGUGCGUUUCUGUGGUAUUGCACUUGUCCCGGCCUGUUGAGGUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001109
          miRBase ID: gga-miR-92-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: NA

          Stem-loop ID gga-mir-92-2
          Stem-loop Accession # MI0029335
          Stem-loop Sequence
          GUUCCUCCGUGGGUGGGGAUUUGUUGCAUUACUUGUAGCUGUGUGUAGAGUAUUGCACUUGUCCCGGCCUGUGGAGGAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001109
          miRBase ID: gga-miR-92-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-92a-1
          Stem-loop Accession # MI0019537
          Stem-loop Sequence
          CAGAAGCGAACCCUUUCUGUGCAGGUUGGGAGAGGUGGCAAUGCUCUGUGCUUGGGGUGGUAUUGCACUUGUCCCGGCCUGUUGAGGACAUGGGGACUGUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022534
          miRBase ID: ola-miR-92a
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: NA

          Stem-loop ID ola-mir-92a-2
          Stem-loop Accession # MI0019419
          Stem-loop Sequence
          UGGAAGAUGUCCUGUCCUGUGCAGGUGGGGAUUAGUAGCAAUGCUGUUCUCCACGAGGUAUUGCACUUGUCCCGGCCUGUAAAGGAACGGGGAGAUGAAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022534
          miRBase ID: ola-miR-92a
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-92a-1
          Stem-loop Accession # MI0004926
          Stem-loop Sequence
          CUUUCUGUAUAGGUUGGGAUUGGUUGCAAUGCUGUACUAUUUAUGUAGUAUUGCACUUGUCCCGGCCUGUUUAGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003580
          miRBase ID: xtr-miR-92a
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: NA

          Stem-loop ID xtr-mir-92a-2
          Stem-loop Accession # MI0004819
          Stem-loop Sequence
          UCCUCUGUGGGUGGGGAUUUGUUGCACUACUGUAUGUAUGAAAAGUAUUGCACUUGUCCCGGCCUGUGGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003580
          miRBase ID: xtr-miR-92a
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn04272657_pri
          TaqMan™ Pri-miRNA Assay : Rn03465594_pri
          TaqMan™ Pri-miRNA Assay : Mm03307329_pri
          TaqMan™ Pri-miRNA Assay : Mm03306814_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10312
          Ambion® Pre-miR™ miRNA Precursor : PM10312
          TaqMan™ MicroRNA Assay : 000430
          mirVana® miRNA inhibitor : MH10312


          miRBase Alias:

          Mouse: mmu-miR-92a (v17)
          Rat: rno-miR-92a (v19)
          Gallus gallus: gga-miR-92 (v19)
          Monodelphis domestica: mdo-miR-92 (v19)
          Xenopus laevis: xla-miR-92a (v21)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-rktn8:80/100.66.72.150:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod