Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480899_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-122-5p
          Stem-loop Accession Number: MI0000442
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 58451074 - 58451158 [+] on Build GRCh38
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Goat, Gorilla gorilla, Guinea pig, Horse, Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pan troglodytes, Paralichthys olivaceus, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480899_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000095, mir-122

          Mature miRNA Sequence:

          UGGAGUGUGACAAUGGUGUUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-122-5p MIMAT0000421
          Mouse mmu-miR-122-5p MIMAT0000246
          Rat rno-miR-122-5p MIMAT0000827
          Alligator mississippiensis ami-miR-122-5p MIMAT0038123
          Anolis carolinensis aca-miR-122-5p MIMAT0021723
          Astatotilapia burtoni abu-miR-122 MIMAT0042117
          View More Rows
          Bovine bta-miR-122 MIMAT0003849
          Callithrix jacchus cja-miR-122 MIMAT0039630
          Chrysemys picta cpi-miR-122-5p MIMAT0037724
          Columba livia cli-miR-122-5p MIMAT0038491
          Dasypus novemcinctus dno-miR-122-5p MIMAT0047610
          Dog cfa-miR-122 MIMAT0006619
          Fugu rubripes fru-miR-122 MIMAT0002991
          Gadus morhua gmo-miR-122-5p MIMAT0044063
          Goat chi-miR-122 MIMAT0035918
          Gorilla gorilla ggo-miR-122 MIMAT0024179
          Guinea pig cpo-miR-122-5p MIMAT0046961
          Horse eca-miR-122 MIMAT0012995
          Metriaclima zebra mze-miR-122 MIMAT0042349
          Neolamprologus brichardi nbr-miR-122 MIMAT0042551
          Oreochromis niloticus oni-miR-122 MIMAT0042727
          Ornithorhynchus anatinus oan-miR-122-5p MIMAT0006825
          Pan troglodytes ptr-miR-122 MIMAT0007963
          Paralichthys olivaceus pol-miR-122-5p MIMAT0025428
          Pongo pygmaeus ppy-miR-122 MIMAT0015746
          Pteropus alecto pal-miR-122-5p MIMAT0039776
          Pundamilia nyererei pny-miR-122 MIMAT0042894
          Python bivittatus pbv-miR-122-5p MIMAT0038909
          Rabbit ocu-miR-122-5p MIMAT0048201
          Rhesus monkey mml-miR-122a-5p MIMAT0006180
          Salmo salar ssa-miR-122-5p MIMAT0032301
          Taeniopygia guttata tgu-miR-122-5p MIMAT0014529
          Tetraodon nigroviridis tni-miR-122 MIMAT0002992
          Zebrafish dre-miR-122 MIMAT0001818

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-122
          Stem-loop Accession # MI0000442
          Stem-loop Sequence
          CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC
          Chromosome Location Chr. 18 - 58451074 - 58451158 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000421
          miRBase ID: hsa-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: Chr. 18 - 58451074 - 58451158 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-122
          Stem-loop Accession # MI0000256
          Stem-loop Sequence
          AGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACCAUCAAACGCCAUUAUCACACUAAAUAGCU
          Chromosome Location Chr. 18 - 65248861 - 65248926 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000246
          miRBase ID: mmu-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: Chr. 18 - 65248861 - 65248926 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-122
          Stem-loop Accession # MI0000891
          Stem-loop Sequence
          CCUUAGCAGAGCUCUGGAGUGUGACAAUGGUGUUUGUGUCCAAAACAUCAAACGCCAUCAUCACACUAAACAGCUACUGCUAGGC
          Chromosome Location Chr. 18 - 60755285 - 60755369 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000827
          miRBase ID: rno-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: Chr. 18 - 60755285 - 60755369 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-122
          Stem-loop Accession # MI0029716
          Stem-loop Sequence
          UACUAUCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038123
          miRBase ID: ami-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-122
          Stem-loop Accession # MI0018719
          Stem-loop Sequence
          UCCUGCUGGAGCUGUGGAGUGUGACAAUGGUGUUUGUAUCCAAUCCGUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021723
          miRBase ID: aca-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-122
          Stem-loop Accession # MI0033823
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUUUCCUCUUCGUCAAACGCCAUUAUCACACUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042117
          miRBase ID: abu-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-122
          Stem-loop Accession # MI0005063
          Stem-loop Sequence
          CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003849
          miRBase ID: bta-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-122
          Stem-loop Accession # MI0032167
          Stem-loop Sequence
          CUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039630
          miRBase ID: cja-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-122
          Stem-loop Accession # MI0029454
          Stem-loop Sequence
          CUGCCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037724
          miRBase ID: cpi-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-122
          Stem-loop Accession # MI0029953
          Stem-loop Sequence
          CUGCCCGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCUGGCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGGUAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038491
          miRBase ID: cli-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-122
          Stem-loop Accession # MI0038963
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCACAAUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047610
          miRBase ID: dno-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-122
          Stem-loop Accession # MI0008015
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006619
          miRBase ID: cfa-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-122
          Stem-loop Accession # MI0003315
          Stem-loop Sequence
          AGCUAUGGAGUGUGACAAUGGUGUUUGUUUCCUUUCUUUCAAACGCCAUUAUCACACUAAAUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002991
          miRBase ID: fru-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-122
          Stem-loop Accession # MI0036011
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUUGUCUGAGCUUUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044063
          miRBase ID: gmo-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-122
          Stem-loop Accession # MI0030600
          Stem-loop Sequence
          CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGGCAAUCCUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035918
          miRBase ID: chi-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-122
          Stem-loop Accession # MI0020726
          Stem-loop Sequence
          GAGUUUCCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGCAAUCCUUCCCUCGAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024179
          miRBase ID: ggo-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-122
          Stem-loop Accession # MI0038621
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046961
          miRBase ID: cpo-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-122
          Stem-loop Accession # MI0012747
          Stem-loop Sequence
          AGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012995
          miRBase ID: eca-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-122
          Stem-loop Accession # MI0034121
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUUUCCUCUUCGUCAAACGCCAUUAUCACACUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042349
          miRBase ID: mze-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-122
          Stem-loop Accession # MI0034406
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUUUCCUCUUCGUCAAACGCCAUUAUCACACUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042551
          miRBase ID: nbr-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-122
          Stem-loop Accession # MI0034648
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUUUCCUCUCCGUCAAACGCCAUUAUCACACUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042727
          miRBase ID: oni-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-122
          Stem-loop Accession # MI0006679
          Stem-loop Sequence
          AACCUUGGAUUGUUUGCUACCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAGUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUAGGGCCAUCCGUCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006825
          miRBase ID: oan-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-122
          Stem-loop Accession # MI0008431
          Stem-loop Sequence
          CUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAAUUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007963
          miRBase ID: ptr-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-122
          Stem-loop Accession # MI0022197
          Stem-loop Sequence
          CUGUGGAGUGUGACAAUGGUGUUUGUGUCCUGUCUAUCAAACGCCAUUAUCACACUAUAUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025428
          miRBase ID: pol-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-122
          Stem-loop Accession # MI0014815
          Stem-loop Sequence
          CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015746
          miRBase ID: ppy-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-122
          Stem-loop Accession # MI0032322
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039776
          miRBase ID: pal-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-122
          Stem-loop Accession # MI0034859
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUUUCCUUUUCGUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042894
          miRBase ID: pny-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-122
          Stem-loop Accession # MI0030210
          Stem-loop Sequence
          UUCUGCUGGAGCUUUGGAGUGUGACAAUGGUGUUUGUAUCCAAUCUCUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038909
          miRBase ID: pbv-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-122
          Stem-loop Accession # MI0039275
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048201
          miRBase ID: ocu-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-122a
          Stem-loop Accession # MI0007613
          Stem-loop Sequence
          CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUACUAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006180
          miRBase ID: mml-miR-122a-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-122-1
          Stem-loop Accession # MI0026450
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCUCUGCUGUCAAACGCCAUUAUUACACUAUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032301
          miRBase ID: ssa-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-122-2
          Stem-loop Accession # MI0026451
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUUCUCUGCAAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032301
          miRBase ID: ssa-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-122
          Stem-loop Accession # MI0013742
          Stem-loop Sequence
          CCCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAUCUAUCAAACGCCAUUAUCACACUAAAUAGUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014529
          miRBase ID: tgu-miR-122-5p
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-122
          Stem-loop Accession # MI0003316
          Stem-loop Sequence
          AGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCUUUCUUUCAAACGCCAUUAUCACACUAAAUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002992
          miRBase ID: tni-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-122
          Stem-loop Accession # MI0001965
          Stem-loop Sequence
          UCCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUAUCAUCUGUCGUCAAACGCCAUUAUCACACUAAAUAGCCACGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001818
          miRBase ID: dre-miR-122
          Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466003_pri
          TaqMan™ Pri-miRNA Assay : Hs03303072_pri
          TaqMan™ Pri-miRNA Assay : Mm03306556_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11012
          Ambion® Pre-miR™ miRNA Precursor : PM11012
          TaqMan™ MicroRNA Assay : 002245
          mirVana® miRNA inhibitor : MH11012
          mirVana® miRNA mimic : MC11012
          TaqMan™ Advanced miRNA Assay : rno480899_mir
          TaqMan™ Advanced miRNA Assay : 477855_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0