Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481430_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-9-3p
          Stem-loop Accession Number: MI0000466
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 156420341 - 156420429 [-] on Build GRCh38
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Species: Human, Mouse, Rat, Monodelphis domestica, Pteropus alecto, Rhesus monkey, Salmo salar, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481430_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000014, mir-9

          Mature miRNA Sequence:

          AUAAAGCUAGAUAACCGAAAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-9-3p MIMAT0000442
          Mouse mmu-miR-9-3p MIMAT0000143
          Rat rno-miR-9a-3p MIMAT0004708
          Monodelphis domestica mdo-miR-9a-3p MIMAT0004088
          Pteropus alecto pal-miR-9-3p MIMAT0039885
          Rhesus monkey mml-miR-9-3-3p MIMAT0031078
          View More Rows
          Salmo salar ssa-miR-9a-4-3p MIMAT0032725
          Salmo salar ssa-miR-9a-5-3p MIMAT0032726
          Taeniopygia guttata tgu-miR-9-3p MIMAT0014627

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-9-1
          Stem-loop Accession # MI0000466
          Stem-loop Sequence
          CGGGGUUGGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGGUGUGGAGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAAUAACCCCA
          Chromosome Location Chr. 1 - 156420341 - 156420429 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000442
          miRBase ID: hsa-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 1 - 156420341 - 156420429 [-] on Build GRCh38

          Stem-loop ID hsa-mir-9-2
          Stem-loop Accession # MI0000467
          Stem-loop Sequence
          GGAAGCGAGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUAUUGGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAACUCCUUCA
          Chromosome Location Chr. 5 - 88666853 - 88666939 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000442
          miRBase ID: hsa-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 1 - 156420341 - 156420429 [-] on Build GRCh38

          Stem-loop ID hsa-mir-9-3
          Stem-loop Accession # MI0000468
          Stem-loop Sequence
          GGAGGCCCGUUUCUCUCUUUGGUUAUCUAGCUGUAUGAGUGCCACAGAGCCGUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGAUUCUCA
          Chromosome Location Chr. 15 - 89368017 - 89368106 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000442
          miRBase ID: hsa-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 1 - 156420341 - 156420429 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-9-1
          Stem-loop Accession # MI0000720
          Stem-loop Sequence
          CGGGGUUGGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGGUGUGGAGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAAUAACCCCA
          Chromosome Location Chr. 3 - 88215598 - 88215686 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000143
          miRBase ID: mmu-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 13 - 83738814 - 83738885 [+] on Build GRCm38

          Stem-loop ID mmu-mir-9-2
          Stem-loop Accession # MI0000157
          Stem-loop Sequence
          GUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUAUUGGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAAC
          Chromosome Location Chr. 13 - 83738814 - 83738885 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000143
          miRBase ID: mmu-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 13 - 83738814 - 83738885 [+] on Build GRCm38

          Stem-loop ID mmu-mir-9-3
          Stem-loop Accession # MI0000721
          Stem-loop Sequence
          GGAGGCCCGUUUCUCUCUUUGGUUAUCUAGCUGUAUGAGUGCCACAGAGCCGUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGACUCUCA
          Chromosome Location Chr. 7 - 79505264 - 79505353 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000143
          miRBase ID: mmu-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 13 - 83738814 - 83738885 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-9a-1
          Stem-loop Accession # MI0000838
          Stem-loop Sequence
          CGGGGUUGGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGGUGUGGAGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAAUAACCCCA
          Chromosome Location Chr. 2 - 187582799 - 187582887 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004708
          miRBase ID: rno-miR-9a-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 2 - 187582799 - 187582887 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-9a-2
          Stem-loop Accession # MI0000840
          Stem-loop Sequence
          GGAAGCGAGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUAUUGGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAACUCCUUCA
          Chromosome Location Chr. 2 - 11884671 - 11884757 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004708
          miRBase ID: rno-miR-9a-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 2 - 187582799 - 187582887 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-9a-3
          Stem-loop Accession # MI0000839
          Stem-loop Sequence
          GGAGGCCCGUUUCUCUCUUUGGUUAUCUAGCUGUAUGAGUGCCACAGAGCCGUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGACUCUAA
          Chromosome Location Chr. 1 - 141229353 - 141229442 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004708
          miRBase ID: rno-miR-9a-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: Chr. 2 - 187582799 - 187582887 [+] on Build Rnor_6.0

          Monodelphis domestica

          Stem-loop ID mdo-mir-9a-1
          Stem-loop Accession # MI0005272
          Stem-loop Sequence
          CGAGGCCCGUUUCUCUCUUUGGUUAUCUAGCUGUAUGAGUGUUAUUGAGCUAUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGACUUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004088
          miRBase ID: mdo-miR-9a-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-9a-2
          Stem-loop Accession # MI0005271
          Stem-loop Sequence
          GGAAGUGAGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUAUUGGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAACUCCUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004088
          miRBase ID: mdo-miR-9a-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-9a-3
          Stem-loop Accession # MI0023762
          Stem-loop Sequence
          UCUUUGGUUAUCUAGCUGUAUGAGUGGUGUCGAGUCUUCAUAAAGCUAGAUAACCGAAAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004088
          miRBase ID: mdo-miR-9a-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-9-1
          Stem-loop Accession # MI0032381
          Stem-loop Sequence
          UCUUUGGUUAUCUAGCUGUAUGAGUGUAUUGGUCUUCAUAAAGCUAGAUAACCGAAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039885
          miRBase ID: pal-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Stem-loop ID pal-mir-9-2
          Stem-loop Accession # MI0032383
          Stem-loop Sequence
          UCUUUGGUUAUCUAGCUGUAUGAGUGCCACAGAGCCGUCAUAAAGCUAGAUAACCGAAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039885
          miRBase ID: pal-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-9-3
          Stem-loop Accession # MI0007586
          Stem-loop Sequence
          GGAGGCCCGUUUCUCUCUUUGGUUAUCUAGCUGUAUGAGUGCCACAGAGCCGUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGACUCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031078
          miRBase ID: mml-miR-9-3-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-9a-4
          Stem-loop Accession # MI0026772
          Stem-loop Sequence
          UCUUUGGUUAUCUAGCUGUAUGAGUGAUGUACAUUCUUCAUAAAGCUAGAUAACCGAAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032725
          miRBase ID: ssa-miR-9a-4-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-9a-5
          Stem-loop Accession # MI0026773
          Stem-loop Sequence
          UCUUUGGUUAUCUAGCUGUAUGAGUGUUAAAUACCUGUCAUAAAGCUAGAUAACCGAAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032726
          miRBase ID: ssa-miR-9a-5-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-9-1
          Stem-loop Accession # MI0013692
          Stem-loop Sequence
          GAUGUUUCUGUCUUUGGUUAUCUAGCUGUAUGAGUGUUGUGGAGCCAUCAUAAAGCUAGAUAACCGAAAGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014627
          miRBase ID: tgu-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA

          Stem-loop ID tgu-mir-9-2
          Stem-loop Accession # MI0013691
          Stem-loop Sequence
          CGAGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUUUUGGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014627
          miRBase ID: tgu-miR-9-3p
          Mature miRNA Sequence: AUAAAGCUAGAUAACCGAAAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303201_pri
          TaqMan™ Pri-miRNA Assay : Mm04227702_pri
          TaqMan™ Pri-miRNA Assay : Mm03307250_pri
          TaqMan™ Pri-miRNA Assay : Rn03464510_pri
          TaqMan™ Pri-miRNA Assay : Rn03465960_pri
          TaqMan™ Pri-miRNA Assay : Rn03465966_pri
          TaqMan™ Pri-miRNA Assay : Hs03303202_pri
          TaqMan™ Pri-miRNA Assay : Mm03306269_pri
          TaqMan™ Pri-miRNA Assay : Hs03293595_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13072
          Ambion® Pre-miR™ miRNA Precursor : PM13072
          TaqMan™ MicroRNA Assay : 002231
          mirVana® miRNA inhibitor : MH13072
          mirVana® miRNA mimic : MC13072
          TaqMan™ Advanced miRNA Assay : 478211_mir
          TaqMan™ Advanced miRNA Assay : rno481430_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline