Search
Search

| Assay Name: | rno-miR-30c-1-3p |
|---|---|
| Stem-loop Accession Number: | MI0000736 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGGGAGAGGGUUGUUUACUCC |
| Species: | Human, Mouse, Rat, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Rabbit |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUGGGAGAGGGUUGUUUACUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30c-1-3p | MIMAT0004674 |
| Mouse | mmu-miR-30c-1-3p | MIMAT0004616 |
| Rat | rno-miR-30c-1-3p | MIMAT0004719 |
| Chrysemys picta | cpi-miR-30c-3p | MIMAT0037679 |
| Cricetulus griseus | cgr-miR-30c | MIMAT0041161 |
| Dasypus novemcinctus | dno-miR-30c-3p | MIMAT0047561 |
| View More Rows | ||
| Rabbit | ocu-miR-30c-3p | MIMAT0048155 |