Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174885.3 | NP_777479.2 | 1 | 1016 | ||
GenBank mRNA | AY158689.1 | - | 1 | 1016 | ||
BC108387.1 | - | 1 | 969 | |||
BC139059.1 | - | 1 | 1001 | 8862081 |