Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirpb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002898.1 | NP_001002898.1 | 3 | 748 | ||
GenBank mRNA | AB109404.1 | - | 825 | |||
AB112022.1 | - | 3 | 748 | |||
AB112023.1 | - | 751 | ||||
AB112024.1 | - | 785 | ||||
AB112025.1 | - | 2 | 331 | |||
AK036935.1 | - | 743 | ||||
AK040982.1 | - | 801 | ||||
AK041165.1 | - | 805 | ||||
AK054545.1 | - | 761 | ||||
AK170671.1 | - | 751 | ||||
BC145417.1 | - | 760 | 9053975 | |||
BC145418.1 | - | 761 | 9053976 |
Gene Symbol: LOC100038947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173459.2 | NP_001166930.1 | 3 | 800 | ||
GenBank mRNA | AK155731.1 | - | 801 | |||
BC039636.1 | - | 744 | ||||
DQ055453.1 | - | 3 | 749 |
Gene Symbol: Sirpb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173460.1 | NP_001166931.1 | 3 | 841 | ||
GenBank mRNA | AK154879.1 | - | 3 | 842 |