Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009392.2 | NP_033418.1 | 3 | 965 | ||
GenBank mRNA | AK013072.1 | - | 2 | 1042 | ||
BC138240.1 | - | 3 | 839 | 8861259 | ||
BC138241.1 | - | 3 | 839 | 8861260 | ||
M75953.1 | - | 3 | 883 |