Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197216.2 | NP_001184145.1 | 3, 4 | 577 | ||
NM_001671.4 | NP_001662.1 | 4, 5 | 694 | |||
XM_011523861.2 | XP_011522163.1 | 4, 5 | 601 | |||
GenBank mRNA | AB070933.1 | - | 473 | |||
AK124286.1 | - | 2, 3 | 1807 | |||
AK308375.1 | - | 4, 5 | 500 | |||
AK308677.1 | - | 2, 3 | 325 | |||
BC032130.2 | - | 4, 5 | 371 | 5087444 | ||
BQ653541.1 | - | 4, 5 | 704 | |||
BT019873.1 | - | 3, 4 | 294 | |||
CR542052.1 | - | 3, 4 | 294 | |||
FJ410292.1 | - | 2, 3 | 177 | |||
M10058.1 | - | 4, 5 | 466 |