Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153151.3 | NP_694791.2 | 8 | 1071 | ||
GenBank mRNA | AF356876.1 | - | 8 | 1077 | ||
AK143884.1 | - | 8 | 1031 | |||
AK149441.1 | - | 8 | 1071 | |||
BC049873.1 | - | 8 | 976 | 5054536 |