Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136068.2 | NP_001129540.1 | 1 | 157 | ||
NM_001288664.1 | NP_001275593.1 | 1 | 157 | |||
NM_010652.2 | NP_034782.1 | 1 | 157 | |||
GenBank mRNA | AF095447.1 | - | 1 | 94 | ||
AF106008.1 | - | 1 | 120 | |||
AF106009.1 | - | 1 | 112 | |||
AF109782.1 | - | 1 | 94 | |||
AF109783.1 | - | 94 | ||||
AF109784.1 | - | 1 | 94 | |||
AK138801.1 | - | 1 | 113 | |||
BC145003.1 | - | 1 | 157 | |||
BC145694.1 | - | 1 | 157 | 40130997 |