Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011353.2 | NP_035483.1 | 2 | 289 | ||
GenBank mRNA | AF073517.1 | - | 2 | 260 | ||
AK005773.1 | - | 2 | 204 | |||
AK012379.1 | - | 2 | 224 | |||
BC028510.1 | - | 2 | 242 | 1510984 | ||
BY102586.1 | - | 2 | 298 |