Gene Symbol: Ly6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135688.1 | NP_001129160.1 | 3, 4 | 394 | ||
NM_001135689.1 | NP_001129161.1 | 3, 4 | 308 | |||
NM_011837.3 | NP_035967.1 | 3, 4 | 671 | |||
GenBank mRNA | AF127091.1 | - | 3, 4 | 304 | ||
AK021002.1 | - | 3, 4 | 348 | |||
AK034884.1 | - | 311 | ||||
BB627076.1 | - | 3, 4 | 311 | |||
BC028758.1 | - | 3, 4 | 305 | 1245940 | ||
BM943584.1 | - | 3, 4 | 394 | |||
BM948855.1 | - | 3, 4 | 671 | |||
CX238172.1 | - | 3, 4 | 374 |