Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 2 | 665 | ||
NM_001290077.1 | NP_001277006.1 | 2 | 665 | |||
XM_005251561.1 | XP_005251618.1 | 2 | 332 | |||
XM_005251563.1 | XP_005251620.1 | 2 | 332 | |||
GenBank mRNA | AB086825.1 | - | 2 | 321 | ||
AB208796.1 | - | 2 | 343 | |||
AK291775.1 | - | 2 | 353 | |||
AK295043.1 | - | 2 | 391 | |||
BC035514.1 | - | 2 | 340 | 5228999 | ||
EU826591.1 | - | 2 | 215 | |||
EU826592.1 | - | 2 | 215 | |||
L06139.1 | - | 2 | 363 |