Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030952.2 | NP_112214.2 | 7 | 1218 | ||
XM_005245515.4 | XP_005245572.1 | 3 | 475 | |||
GenBank mRNA | AK074830.1 | - | 7 | 1172 | ||
AK303577.1 | - | 7 | 1218 | |||
AL136891.1 | - | 7 | 1182 | |||
BC017306.2 | - | 7 | 1154 | 5088037 | ||
BX537564.1 | - | 1 | 4255 | |||
CR749209.1 | - | 3 | 4647 |