Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251877.1 | NP_001238806.1 | 5, 6 | 714 | ||
NM_003363.3 | NP_003354.2 | 5, 6 | 714 | |||
NM_199443.2 | NP_955475.1 | 5, 6 | 714 | |||
GenBank mRNA | AF017305.1 | - | 5, 6 | 638 | ||
AF017306.1 | - | 5, 6 | 635 | |||
AK222725.1 | - | 5, 6 | 644 | |||
AK223292.1 | - | 5, 6 | 644 | |||
AK291795.1 | - | 5, 6 | 659 | |||
BC125130.1 | - | 5, 6 | 685 | 40119090 | ||
BC125131.1 | - | 5, 6 | 682 | 40119091 | ||
U20657.1 | - | 5, 6 | 638 |