Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128610.2 | NP_001122082.1 | 7, 8 | 1036 | ||
NM_001283049.1 | NP_001269978.1 | 5, 6 | 665 | |||
NM_005154.4 | NP_005145.3 | 7, 8 | 896 | |||
XM_006720761.3 | XP_006720824.1 | 8, 9 | 1216 | |||
XM_006720762.3 | XP_006720825.1 | 8, 9 | 1216 | |||
XM_011522193.2 | XP_011520495.1 | 8, 9 | 1053 | |||
XM_017022718.1 | XP_016878207.1 | 7, 8 | 891 | |||
XM_017022719.1 | XP_016878208.1 | 7, 8 | 1008 | |||
XM_017022720.1 | XP_016878209.1 | 8, 9 | 1053 | |||
XM_017022721.1 | XP_016878210.1 | 8, 9 | 953 | |||
XM_017022722.1 | XP_016878211.1 | 6, 7 | 876 | |||
GenBank mRNA | AK292400.1 | - | 7, 8 | 891 | ||
AK296480.1 | - | 5, 6 | 655 | |||
BC015545.1 | - | 8, 9 | 1170 | |||
BC026954.1 | - | 7, 8 | 986 | |||
BC038801.1 | - | 7, 8 | 884 | |||
BC051345.1 | - | 7, 8 | 935 | |||
BC092443.1 | - | 6, 7 | 695 | |||
BC110590.2 | - | 7, 8 | 862 | 40027270 | ||
BX537420.1 | - | 7, 8 | 891 | |||
D29956.1 | - | 7, 8 | 1010 |