Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006741.3 | NP_006732.3 | 7 | 1385 | ||
GenBank mRNA | BC022470.1 | - | 7 | 1385 | 4819907 |