Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004523.3 | NP_004514.2 | 16 | 2439 | ||
GenBank mRNA | AB208964.1 | - | 2 | 174 | ||
AK314258.1 | - | 16 | 2342 | |||
BC126211.1 | - | 16 | 2238 | 8991927 | ||
BC136474.1 | - | 16 | 2238 | 9020463 | ||
U37426.1 | - | 16 | 2221 | |||
X85137.1 | - | 16 | 2144 |