Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021643.3 | NP_067675.1 | 3 | 3577 | ||
NR_027303.1 | 3 | 2010 | ||||
GenBank mRNA | AK129973.1 | - | 1 | 1235 | ||
D87119.1 | - | 3 | 3375 |